missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-3-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. Q00801G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q00801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q00801G Supplier TCI America Supplier No. Q00801G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 191162-39-7
Molecular Formula C9H8BNO2
Molecular Weight (g/mol) 172.98
MDL Number MFCD02183527
InChI Key YGDICLRMNDWZAK-UHFFFAOYSA-N
PubChem CID 2734663
IUPAC Name (quinolin-3-yl)boronic acid
SMILES OB(O)C1=CC2=CC=CC=C2N=C1

Specifications

Quantity 1 g
Synonym 3-quinolineboronic acid, quinoline-3-boronic acid, 3-quinolinylboronic acid, 3-quinolylboronic acid, quinolin-3-yl-3-boronic acid, 3-quinoline boronic acid, 3-quinolineboronic aid, quinolin-3-yl-boranediol, quinolin-3-yl boronic acid, boronic acid, 3-quinolinyl
Formula Weight 172.98
Physical Form Crystalline Powder
Chemical Name or Material Quinoline-3-boronic Acid (contains varying amounts of Anhydride)