missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-3-boronic acid, 97%

Catalog No. p-4510684
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC367730010 1 g Glass bottle
AC367730050 5 g Glass bottle
AC367730250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC367730010 Supplier Thermo Scientific Chemicals Supplier No. 367730010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 191162-39-7 | C9H8BNO2 | 172.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 191162-39-7
Molecular Formula C9H8BNO2
Molecular Weight (g/mol) 172.98
MDL Number MFCD02183527
InChI Key YGDICLRMNDWZAK-UHFFFAOYSA-N
Synonym 3-quinolineboronic acid, quinoline-3-boronic acid, 3-quinolinylboronic acid, 3-quinolylboronic acid, quinolin-3-yl-3-boronic acid, 3-quinoline boronic acid, 3-quinolineboronic aid, quinolin-3-yl-boranediol, quinolin-3-yl boronic acid, boronic acid, 3-quinolinyl
PubChem CID 2734663
IUPAC Name quinolin-3-ylboronic acid
SMILES OB(O)C1=CC2=CC=CC=C2N=C1

Specifications

Melting Point 148.0°C to 155.0°C
Color Brown to Orange or Pink
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 172.98
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Quinoline-3-boronic acid
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only