missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-3-boronic acid, 95%

CAS: 191162-39-7 | C9H8BNO2 | 172.98 g/mol

$74.67 - $285.99

Chemical Identifiers

CAS 191162-39-7
Molecular Formula C9H8BNO2
Molecular Weight (g/mol) 172.98
MDL Number MFCD02183527
InChI Key YGDICLRMNDWZAK-UHFFFAOYSA-N
Synonym 3-quinolineboronic acid, quinoline-3-boronic acid, 3-quinolinylboronic acid, 3-quinolylboronic acid, quinolin-3-yl-3-boronic acid, 3-quinoline boronic acid, 3-quinolineboronic aid, quinolin-3-yl-boranediol, quinolin-3-yl boronic acid, boronic acid, 3-quinolinyl
PubChem CID 2734663
SMILES OB(O)C1=CC2=CC=CC=C2N=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL20088MD
View Documents
Thermo Scientific Chemicals
L20088MD
250 mg
Each for $74.67
Only null left
 
AAL2008803
View Documents
Thermo Scientific Chemicals
L2008803
1 g
Each for $85.62
Only null left
 
AAL2008806
View Documents
Thermo Scientific Chemicals
L2008806
5 g
Each for $285.99
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

191162-39-7
172.98
YGDICLRMNDWZAK-UHFFFAOYSA-N
2734663
C9H8BNO2
MFCD02183527
3-quinolineboronic acid, quinoline-3-boronic acid, 3-quinolinylboronic acid, 3-quinolylboronic acid, quinolin-3-yl-3-boronic acid, 3-quinoline boronic acid, 3-quinolineboronic aid, quinolin-3-yl-boranediol, quinolin-3-yl boronic acid, boronic acid, 3-quinolinyl
OB(O)C1=CC2=CC=CC=C2N=C1

Specifications

191162-39-7
C9H8BNO2
250 mg
3-quinolineboronic acid, quinoline-3-boronic acid, 3-quinolinylboronic acid, 3-quinolylboronic acid, quinolin-3-yl-3-boronic acid, 3-quinoline boronic acid, 3-quinolineboronic aid, quinolin-3-yl-boranediol, quinolin-3-yl boronic acid, boronic acid, 3-quinolinyl
OB(O)C1=CC2=CC=CC=C2N=C1
172.98
172.98
Quinoline-3-boronic acid
>300°C
MFCD02183527
8764547
YGDICLRMNDWZAK-UHFFFAOYSA-N
quinolin-3-ylboronic acid
2734663
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only