missing translation for 'onlineSavingsMsg'
Learn More

Quinine monohydrochloride dihydrate, 99% (total base), may contain up to 10% Dihydroquinine

CAS: 6119-47-7 | C20H29ClN2O4 | 396.91 g/mol

$82.62 - $252.44

Chemical Identifiers

CAS 6119-47-7
Molecular Formula C20H29ClN2O4
Molecular Weight (g/mol) 396.91
MDL Number MFCD00151248
InChI Key MPQKYZPYCSTMEI-VQYSEXPNNA-N
Synonym quinine hcl dihydrate
PubChem CID 124080947
IUPAC Name (R)-[(2S,4S,5S)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;dihydrate;hydrochloride
SMILES O.O.Cl.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3347409
View Documents
Thermo Scientific Chemicals
H3347409
10 g
Each for $82.62
Only null left
 
AAH3347418
View Documents
Thermo Scientific Chemicals
H3347418
50 g
Each for $252.44
Only null left
 
Description

Description

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

6119-47-7
396.91
MPQKYZPYCSTMEI-VQYSEXPNNA-N
124080947
O.O.Cl.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C
C20H29ClN2O4
MFCD00151248
quinine hcl dihydrate
(R)-[(2S,4S,5S)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;dihydrate;hydrochloride

Specifications

6119-47-7
MFCD00151248
6112655
14,8061
Sparingly soluble in water.(0.26g/L) (25°C),
−246° (c=2 in 0.1M HCl)
(R)-[(2S,4S,5S)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;dihydrate;hydrochloride
124080947
99%
(total base), may contain up to 10% Dihydroquinine
C20H29ClN2O4
10 g
Light sensitive
quinine hcl dihydrate
MPQKYZPYCSTMEI-VQYSEXPNNA-N
O.O.Cl.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C
396.91
396.91 (360.88 Anhydrous)
Quinine monohydrochloride dihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c

H302-H317-H335

EINECSNumber : 205-001-1

RTECSNumber : GD2947000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only