missing translation for 'onlineSavingsMsg'
Learn More

Quinine monohydrochloride dihydrate, 99% (total base), may contain up to 10% Dihydroquinine

Catalog No. AAH3347409
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3347409 10 g
AAH3347418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3347409 Supplier Thermo Scientific Chemicals Supplier No. H3347409

CAS: 6119-47-7 | C20H29ClN2O4 | 396.91 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 6119-47-7
Molecular Formula C20H29ClN2O4
Molecular Weight (g/mol) 396.91
MDL Number MFCD00151248
InChI Key MPQKYZPYCSTMEI-VQYSEXPNNA-N
Synonym quinine hcl dihydrate
PubChem CID 124080947
IUPAC Name (R)-[(2S,4S,5S)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;dihydrate;hydrochloride
SMILES O.O.Cl.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C

Specifications

Quantity 10 g
Beilstein 6112655
Sensitivity Light sensitive
Merck Index 14,8061
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Optical Rotation −246° (c=2 in 0.1M HCl)
Formula Weight 396.91 (360.88 Anhydrous)
Percent Purity 99%
Assay (total base), may contain up to 10% Dihydroquinine
Chemical Name or Material Quinine monohydrochloride dihydrate
Hazard Category H302-H317-H335
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
EINECSNumber 205-001-1
RTECSNumber GD2947000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only