missing translation for 'onlineSavingsMsg'
Learn More

Quinidine 85.0+%, TCI America™

Supplier:  TCI America Q000625G

 View more versions of this product

Catalog No. Q000625G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Quinidine
172°C
C20H24N2O2
25 g
quinidine, +-quinidine, chinidin, pitayine, cin-quin, 8r,9s-quinidine, conquinine, conchinin, quinidex, conchinine
COC1=CC=C2N=CC=C([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)C2=C1
324.42
CHEBI:28593
≥85.0% (HPLC)
56-54-2
White
MFCD00135581,MFCD00198096
2811
LOUPRKONTZGTKE-LHHVKLHASA-N
(S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol
441074
324.42
Crystalline Powder

Chemical Identifiers

56-54-2
324.42
LOUPRKONTZGTKE-LHHVKLHASA-N
441074
(S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol
C20H24N2O2
MFCD00135581,MFCD00198096
quinidine, +-quinidine, chinidin, pitayine, cin-quin, 8r,9s-quinidine, conquinine, conchinin, quinidex, conchinine
CHEBI:28593
COC1=CC=C2N=CC=C([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)C2=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0324

RTECSNumber : VA4725000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions