missing translation for 'onlineSavingsMsg'
Learn More

Quinidine 85.0+%, TCI America™

Catalog No. p-7135614
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
Q00065G 5 g
Q000625G 25 g
2 options

Catalog No. Q00065G

Supplier: TCI America Q00065G

Only null left

Chemical Identifiers

CAS 56-54-2
Molecular Formula C20H24N2O2
Molecular Weight (g/mol) 324.42
MDL Number MFCD00135581,MFCD00198096
InChI Key LOUPRKONTZGTKE-LHHVKLHASA-N
Synonym quinidine, +-quinidine, chinidin, pitayine, cin-quin, 8r,9s-quinidine, conquinine, conchinin, quinidex, conchinine
PubChem CID 441074
ChEBI CHEBI:28593
IUPAC Name (S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol
SMILES COC1=CC=C2N=CC=C([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)C2=C1

Specifications

Melting Point 172°C
Color White
Quantity 5 g
UN Number 2811
Formula Weight 324.42
Percent Purity ≥85.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Quinidine
EINECSNumber (9)-0324
RTECSNumber VA4725000
TSCA Yes
Recommended Storage Refrigerator