missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Quinic acid, MilliporeSigma™ Supelco™
Description
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| Packaging | Ampule |
| CAS | 77-95-2 |
| Physical Form | Neat |
| Linear Formula | C7H12O6 |
| MDL Number | MFCD00003864 |
| Synonym | D-(-)-Quinic acid |
| Type | Food and Cosmetic Component Standard |
| Quantity | 500 mg |
| Formula Weight | 192.17 |
| Grade | Analytical Standard |