missing translation for 'onlineSavingsMsg'
Learn More

Quincoridine 95.0+%, TCI America™

Catalog No. Q0076100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q0076100MG 100 mg
Q00761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q0076100MG Supplier TCI America Supplier No. Q0076100MG

Chemical Identifiers

CAS 207129-36-0
Molecular Formula C10H17NO
Molecular Weight (g/mol) 167.252
InChI Key GAFZBOMPQVRGKU-UDNWOFFPSA-N
Synonym (1S,2R,5R)-2-(Hydroxymethyl)-5-vinylquinuclidine, (1S,2R,5R)-5-Vinyl-2-quinuclidinemethanol
PubChem CID 24058276
IUPAC Name [(2R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]methanol
SMILES C=CC1CN2CCC1CC2CO

Specifications

Color Yellow
Boiling Point 267°C
Quantity 100 mg
Formula Weight 167.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Quincoridine