missing translation for 'onlineSavingsMsg'
Learn More

Quinazoline 98.0+%, TCI America™

Catalog No. q00551g
Change view
Click to view available options
Quantity:
1 g
Catalog No. Quantity
Q00551G 1 g
1 options

Catalog No. Q00551G

Supplier: TCI America Q00551G

Only null left

Chemical Identifiers

CAS 253-82-7
Molecular Formula C8H6N2
Molecular Weight (g/mol) 130.15
MDL Number MFCD00006712
InChI Key JWVCLYRUEFBMGU-UHFFFAOYSA-N
Synonym phenmiazine, 1,3-benzodiazine, 1,3-diazanaphthalene, 5,6-benzopyrimidine, benzo a pyrimidine, chinazolin, unii-ub9qur18nl, ub9qur18nl, benzopyrimidine
PubChem CID 9210
ChEBI CHEBI:36621
IUPAC Name quinazoline
SMILES C1=CC=C2N=CN=CC2=C1

Specifications

Melting Point 48°C
Boiling Point 243°C
Quantity 1 g
Formula Weight 130.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Quinazoline