CAS RN 253-82-7
IUPAC Name:
quinazoline
Synonyms:
quinazoline benzo(a)pyrimidine phenmiazine 5,6-benzopyrimidine chinazolin benzo[a]pyrimidine
Molecular Weight (g/mol):
130.15
Molecular Formula:
C8H6N2
InChi Key:
JWVCLYRUEFBMGU-UHFFFAOYSA-N
SMILES:
C1=CC=C2N=CN=CC2=C1
1
–
1
of
1
results
1
Quinazoline 98.0+%, TCI America™
CAS: 253-82-7 Molecular Formula: C8H6N2 Molecular Weight (g/mol): 130.15 MDL Number: MFCD00006712 InChI Key: JWVCLYRUEFBMGU-UHFFFAOYSA-N Synonym: phenmiazine,1,3-benzodiazine,1,3-diazanaphthalene,5,6-benzopyrimidine,benzo a pyrimidine,chinazolin,unii-ub9qur18nl,ub9qur18nl,benzopyrimidine PubChem CID: 9210 ChEBI: CHEBI:36621 IUPAC Name: quinazoline SMILES: C1=CC=C2N=CN=CC2=C1