missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Quinaldine red, pure

CAS: 117-92-0 | C21H23IN2 | 430.33 g/mol

$129.02

Chemical Identifiers

CAS 117-92-0
Molecular Formula C21H23IN2
Molecular Weight (g/mol) 430.33
MDL Number MFCD00011968
InChI Key JOLANDVPGMEGLK-UHFFFAOYSA-M
Synonym 2-[4-(Dimethylamino)styryl]-1-ethylquinolinium iodide
PubChem CID 5702759
SMILES [I-].CC[N+]1=C2C=CC=CC2=CC=C1C=CC1=CC=C(C=C1)N(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC377510050
View Documents Promotion Details
Thermo Scientific Chemicals
377510050
5 g Glass bottle
Each for $129.02
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

117-92-0
430.33
JOLANDVPGMEGLK-UHFFFAOYSA-M
5702759
C21H23IN2
MFCD00011968
2-[4-(Dimethylamino)styryl]-1-ethylquinolinium iodide
[I-].CC[N+]1=C2C=CC=CC2=CC=C1C=CC1=CC=C(C=C1)N(C)C

Specifications

117-92-0
MFCD00011968
JOLANDVPGMEGLK-UHFFFAOYSA-M
Authentic
430.33
430.32
Complies
Powder
C21H23IN2
2-[4-(Dimethylamino)styryl]-1-ethylquinolinium iodide
[I-].CC[N+]1=C2C=CC=CC2=CC=C1C=CC1=CC=C(C=C1)N(C)C
2-{2-[4-(dimethylamino)phenyl]ethenyl}-1-ethylquinolin-1-ium iodide
5702759
Pure
Black/Green or Blue to Gray
Quinaldine red, Pure
Videos
SDS
Documents

Documents

RUO – Research Use Only