missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Quinaldine red, pure

Catalog No. AC377510050
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC377510050 5 g Glass bottle
AC377511000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC377510050 Supplier Thermo Scientific Chemicals Supplier No. 377510050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 117-92-0 | C21H23IN2 | 430.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 117-92-0
Molecular Formula C21H23IN2
Molecular Weight (g/mol) 430.33
MDL Number MFCD00011968
InChI Key JOLANDVPGMEGLK-UHFFFAOYSA-M
Synonym 2-[4-(Dimethylamino)styryl]-1-ethylquinolinium iodide
PubChem CID 5702759
SMILES [I-].CC[N+]1=C2C=CC=CC2=CC=C1C=CC1=CC=C(C=C1)N(C)C

Specifications

Infrared Spectrum Authentic
IUPAC Name 2-{2-[4-(dimethylamino)phenyl]ethenyl}-1-ethylquinolin-1-ium iodide
Formula Weight 430.32
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Black/Green or Blue to Gray
Quantity 5 g
Physical Form Powder
Chemical Name or Material Quinaldine red, Pure

RUO – Research Use Only