missing translation for 'onlineSavingsMsg'
Learn More

Quinaldine Ethiodide 99.0+%, TCI America™

Catalog No. E017225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E017225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E017225G Supplier TCI America Supplier No. E017225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 606-55-3
Molecular Formula C12H14IN
Molecular Weight (g/mol) 299.155
MDL Number MFCD00031829
InChI Key OEVSHJVOKFWBJY-UHFFFAOYSA-M
Synonym 1-ethyl-2-methylquinolinium iodide, quinaldine ethiodide, 1-ethylquinaldinium iodide, n-ethylquinaldinium iodide, 2-methylquinoline ethiodide, quinaldinium, 1-ethyl-, iodide, quinolinium, 1-ethyl-2-methyl-, iodide, 2-methyl-1-ethylquinolinium iodide, 1-ethyl-2-methylquinolin-1-ium iodide, quinolinium, 1-ethyl-2-methyl-, iodide 1:1
PubChem CID 69076
IUPAC Name 1-ethyl-2-methylquinolin-1-ium;iodide
SMILES CC[N+]1=C(C=CC2=CC=CC=C21)C.[I-]

Specifications

Quantity 25 g
Formula Weight 299.16
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Quinaldine Ethiodide
RTECSNumber VA0740000
TSCA Yes