accessibility menu, dialog, popup

UserName

IH3CNH3C

CAS RN 606-55-3

IUPAC Name: 1-ethyl-2-methylquinolin-1-ium iodide
Synonyms: 1-ethyl-2-methylquinolin-1-ium iodide
Molecular Weight (g/mol): 299.16
Molecular Formula: C12H14IN
InChi Key: OEVSHJVOKFWBJY-UHFFFAOYSA-M
SMILES: [I-].CC[N+]1=C2C=CC=CC2=CC=C1C
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)