Learn More
Quinaldine, 96%
CAS: 91-63-4 | C10H9N | 143.19 g/mol
$50.67 - $448.38
Chemical Identifiers
| CAS | 91-63-4 |
|---|---|
| Molecular Formula | C10H9N |
| Molecular Weight (g/mol) | 143.19 |
| MDL Number | MFCD00006756 |
| InChI Key | SMUQFGGVLNAIOZ-UHFFFAOYSA-N |
| Synonym | quinaldine, chinaldine, khinaldin, quinoline, 2-methyl, 2-methyl-quinoline, methylquinoline, 2-methylchinolin, ccris 1155, 2-methylchinoline, 2-methylchinolin czech |
| PubChem CID | 7060 |
| IUPAC Name | 2-methylquinoline |
| SMILES | CC1=CC=C2C=CC=CC2=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1341214
|
Thermo Scientific Chemicals
A1341214 |
25 g |
Each for $50.67
|
|
|||||
|
AAA1341222
|
Thermo Scientific Chemicals
A1341222 |
100 g |
Each for $136.64
|
|
|||||
|
AAA1341236
|
Thermo Scientific Chemicals
A1341236 |
500 g |
Each for $448.38
|
|
|||||
Description
It is an anaesthetic used in fish transportation. Used in preparation of oil-soluble dyes. Also used in anti-malaria drugs, in manufacturing dyes, food colorants , pharmaceuticals and pH indicators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an anaesthetic used in fish transportation. Used in preparation of oil-soluble dyes. Also used in anti-malaria drugs, in manufacturing dyes, food colorants , pharmaceuticals and pH indicators.
Solubility
Soluble in water (2.5g/L).
Notes
Air and Light Sensitive. Store in dark and away from air. Store in cool, dry place in tightly closed container. Store away from acid chlorides and acid anhydrides. It may discolor to brown upon storage.
Chemical Identifiers
| 91-63-4 | |
| 143.19 | |
| SMUQFGGVLNAIOZ-UHFFFAOYSA-N | |
| 7060 | |
| CC1=CC=C2C=CC=CC2=N1 |
| C10H9N | |
| MFCD00006756 | |
| quinaldine, chinaldine, khinaldin, quinoline, 2-methyl, 2-methyl-quinoline, methylquinoline, 2-methylchinolin, ccris 1155, 2-methylchinoline, 2-methylchinolin czech | |
| 2-methylquinoline |
Specifications
| 91-63-4 | |
| 1.058 | |
| 79°C (174°F) | |
| C10H9N | |
| MFCD00006756 | |
| 110309 | |
| 14,8047 | |
| Soluble in water (2.5g/L). | |
| CC1=CC=C2C=CC=CC2=N1 | |
| 143.19 | |
| 143.19 | |
| Quinaldine |
| -2°C | |
| 247°C to 248°C | |
| Odorless | |
| 1.612 | |
| 25 g | |
| Air and light sensitive | |
| quinaldine, chinaldine, khinaldin, quinoline, 2-methyl, 2-methyl-quinoline, methylquinoline, 2-methylchinolin, ccris 1155, 2-methylchinoline, 2-methylchinolin czech | |
| SMUQFGGVLNAIOZ-UHFFFAOYSA-N | |
| 2-methylquinoline | |
| 7060 | |
| ≥97% |
Safety and Handling
GHS H Statement
H302-H312-H315-H319-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P201-P202-P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P374-P501c
H227-H302+H312-H315-H319-H335-H341
EINECSNumber : 202-085-1
RTECSNumber : UZ9625000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only