missing translation for 'onlineSavingsMsg'
Learn More

Quinaldic Acid Sodium Salt 98.0+%, TCI America™

Catalog No. p-7135612
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q00041G 1 g
Q000425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q00041G Supplier TCI America Supplier No. Q00041G
Only null left

Chemical Identifiers

CAS 16907-79-2
Molecular Formula C10H6NNaO2
Molecular Weight (g/mol) 195.153
MDL Number MFCD00070526
InChI Key PLTCLMZAIZEHGD-UHFFFAOYSA-M
Synonym Sodium Quinaldinate, Sodium 2-Quinolinecarboxylate, 2-Quinolinecarboxylic Acid Sodium Salt
PubChem CID 23672094
IUPAC Name sodium;quinoline-2-carboxylate
SMILES C1=CC=C2C(=C1)C=CC(=N2)C(=O)[O-].[Na+]

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 195.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Quinaldic Acid Sodium Salt