accessibility menu, dialog, popup

UserName

NaOHON

CAS RN 16907-79-2

IUPAC Name: quinoline-2-carboxylic acid sodium
Synonyms: quinaldic acid sodium
Molecular Weight (g/mol): 196.16
Molecular Formula: C10H7NNaO2
InChi Key: RNUUPIHGELIDIL-UHFFFAOYSA-N
SMILES: [Na].OC(=O)C1=CC=C2C=CC=CC2=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results