missing translation for 'onlineSavingsMsg'
Learn More

QUIN 2, Tetrapotassium Salt,, MilliporeSigma™

Catalog No. 55182650mg
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55182650MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55182650MG Supplier MilliporeSigma Supplier No. 55182650MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Binding of Ca2+ leads to a major shift in UV absorption spectrum and a twenty-fold enhancement in fluorescence quantum yield

Chemical Identifiers

CAS 73630-23-6
Molecular Formula C26H23K4N3O10
Molecular Weight (g/mol) 693.87
MDL Number MFCD00065487
InChI Key XAGUNWDMROKIFJ-UHFFFAOYSA-J
Synonym quin 2, quin2, quin 2,tetrapotassium salt, unii-o448idk23o, quin 2, tetrapotassium salt, glycine, n-2-8-bis carboxymethyl amino-6-methoxy-2-quinolinyl methoxy-4-methylphenyl-n-carboxymethyl-, tetrapotassium salt, tetrapotassium 2-8-bis carboxylatomethyl amino-6-methoxyquinolin-2-yl methoxy-4-methylphenyl carboxylatomethyl amino acetate, quin-2, suitable for fluorescence hplc, tetrapotassium 2-2-8-bis 2-oxido-2-oxoethyl amino-6-methoxyquinolin-2-yl methoxy-4-methyl-n-2-oxido-2-oxoethyl anilino acetate
PubChem CID 135539
IUPAC Name tetrapotassium 2-{[2-({2-[bis(carboxylatomethyl)amino]-5-methylphenoxy}methyl)-6-methoxyquinolin-8-yl](carboxylatomethyl)amino}acetate
SMILES [K+].[K+].[K+].[K+].COC1=CC(N(CC([O-])=O)CC([O-])=O)=C2N=C(COC3=CC(C)=CC=C3N(CC([O-])=O)CC([O-])=O)C=CC2=C1

Specifications

Color Yellow
Quantity 50 mg
Formula Weight 693.9
Percent Purity ≥95%
Grade HPLC
Physical Form Solid
Chemical Name or Material QUIN 2, Tetrapotassium Salt
Hazard Category H315: Causes skin irritation.
Hazard Statement H319: Causes serious eye irritation.
Precautionary Statement H335: May cause respiratory irritation
Recommended Storage -20°C (-4°F)