accessibility menu, dialog, popup

UserName

KKKKCH3OOH3CNOOOONNOOOO

CAS RN 73630-23-6

IUPAC Name: tetrapotassium 2-{[2-({2-[bis(carboxylatomethyl)amino]-5-methylphenoxy}methyl)-6-methoxyquinolin-8-yl](carboxylatomethyl)amino}acetate
Synonyms: tetrapotassium 2-[(2-{2-[bis(carboxylatomethyl)amino]-5-methylphenoxymethyl}-6-methoxyquinolin-8-yl)(carboxylatomethyl)amino]acetate
Molecular Weight (g/mol): 693.87
Molecular Formula: C26H23K4N3O10
InChi Key: XAGUNWDMROKIFJ-UHFFFAOYSA-J
SMILES: [K+].[K+].[K+].[K+].COC1=CC(N(CC([O-])=O)CC([O-])=O)=C2N=C(COC3=CC(C)=CC=C3N(CC([O-])=O)CC([O-])=O)C=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results