missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ QuikChIP (with Loading Control) Kit (ChIP)

Catalog No. NBP229497
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229497 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP229497 Supplier R&D Systems™ Supplier No. NBP229497

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Applications: Chromatin Immunoprecipitation

Specifications

Target QuikChIP (with Loading Control)
For Use With (Application) Western Blotting
Product Type Kit (ChIP)
Content And Storage Wash Buffer A 25ml, Wash Buffer B 25ml, Wash Buffer C 25ml, Wash Buffer D 25ml, SDS Lysis Buffer 50ml, 10X Glycine 25ml, 10X PBS 55ml, ChIP Dilution Buffer 50ml, Salmon Sperm DNA/ Protein A/G Agarose 2 x 1.75mL, RNA Pol II Antibody 1mg/ml 25μL, RNAse A 10 mg/ml 60μL, Human GAPDH Primers 10 pmole/μL 100μL, Negative Control Primers 10 pmole/μL 100μL, 100X PMSF 0.75ml, 100X PIC 0.75mL, Proteinase K (10 mg/ml) 60μL, 20% SDS 625μL, 1M NaHCO3 1.25ml, 0.5M EDTA 250μL, 5M NaCl 500μL, 1M Tris-HCl pH 6.5 500μL. Storage of components varies. See protocol for specific instructions.
Product Line Novus
Quantity 1 Kit