missing translation for 'onlineSavingsMsg'
Learn More

Quidel QuickVue™ Dipstick Strep A Test

Detects Group A Streptococcal antigen directly from patient throat swab specimens or confirms presumptive Group A Streptococcal colonies recovered from culture. Used as an aid in the diagnosis of Group A Streptococcal infection. Lateral flow immunoassay, using Quidel's patented assay device.

Specifications

Content And Storage Room temperature for up to 24 months after date of manufacture.
CPT Code 87880, 87880QW
Clia Complexity Waived
Detectable Analytes Streptococci (Group A) Antigen
Sample Type Throat Swab
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
361042474
View Documents Promotion Details
Quidel
Q20125
25 Tests/Kit N/A
N/A
23043051
View Documents Promotion Details
Quidel
Q20108IN
50 Tests/Kit N/A
N/A
Description

Description

  • CLIA Complexity: Waived
  • CPT Codes: 87880/87880QW
  • Sensitivity: 92%
  • Specificity: 98%
  • Overall Accuracy: 95%
  • Time to Results: Read at five minutes
  • Specimen Type: Throat Swab
  • Internal Procedure Controls: Positive and negative
  • External Controls: Positive and negative liquid

Patent No: 5770460 (June 2015)

Specifications

Specifications

Room temperature for up to 24 months after date of manufacture.
Waived
Throat Swab
5 min.
87880, 87880QW
Streptococci (Group A) Antigen
Dipstick Streptococcus A Test
Videos
SDS
Documents

Documents