missing translation for 'onlineSavingsMsg'
Learn More

Quidel QuickVue™ Dipstick Strep A Test

Catalog No. 361042474
Change view
Click to view available options
Quantity:
25 Tests/Kit
50 Tests/Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361042474 25 Tests/Kit
23043051 50 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 361042474 Supplier Quidel Supplier No. Q20125
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Detects Group A Streptococcal antigen directly from patient throat swab specimens or confirms presumptive Group A Streptococcal colonies recovered from culture. Used as an aid in the diagnosis of Group A Streptococcal infection. Lateral flow immunoassay, using Quidel's patented assay device.

  • CLIA Complexity: Waived
  • CPT Codes: 87880/87880QW
  • Sensitivity: 92%
  • Specificity: 98%
  • Overall Accuracy: 95%
  • Time to Results: Read at five minutes
  • Specimen Type: Throat Swab
  • Internal Procedure Controls: Positive and negative
  • External Controls: Positive and negative liquid

Patent No: 5770460 (June 2015)

Specifications

Content And Storage Room temperature for up to 24 months after date of manufacture.
CPT Code 87880, 87880QW
Clia Complexity Waived
Detectable Analytes Streptococci (Group A) Antigen
Sample Type Throat Swab
Type Dipstick Streptococcus A Test
Quantity 25 Tests/Kit
Test Time 5 min.