Learn More
New Pig Quick Deploy Oil - Only Absorbent Boom
Create a low-profile absorbent barrier for rapid response to oil spills on land. Absorbs and retains oils and oil-based liquids - including lubricants and fuels
Supplier:  New Pig BOM1200
Catalog No. 17-987-825
Only null left
Explore available promotions
Description
Description
  • Quickly contains and absorbs oil spills on land: The boom is designed for rapid deployment to create a low-profile absorbent barrier, preventing the spread of oil.
  • Absorbs and retains oils: It effectively soaks up and holds onto oils, lubricants, and fuels, ensuring they are contained and removed from the environment.
  • Oil-only absorbent: Specifically engineered to absorb oil-based liquids while repelling water, making it highly efficient for spills in wet conditions.
  • Allows clean water to pass through: The specialized material lets clean water filter through, preventing the boom from becoming saturated with water and maximizing its oil-absorption capacity.
  • Durable and reliable: Manufactured by New Pig, a trusted supplier, this boom is built to withstand harsh conditions and provide a reliable solution for spill response.
  • Low-profile design: Its compact shape allows for easy placement and creates a discreet barrier that is less obtrusive than traditional booms.
Product Suggestions
Product Suggestions