missing translation for 'onlineSavingsMsg'
Learn More

Quetiapine Fumarate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018680
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018680 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018680 Supplier Merck Emd Millipore Supplier No. PHR18561G

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 111974-72-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C21H25N3O2 S · 0.5C4H4O4
MDL Number MFCD03423782
Synonym Quetiapine hemifumarate salt; 2-[2-(4-Dibenzo[b,f][1,4 ]thiazepin-11-yl-1-piperazinyl)ethoxy]ethanol hemifumarate; Quetiapine fumarate
Type Certified Reference Material
Quantity 1 g
Formula Weight 441.54
Grade Certified Reference Material