missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quercetin 3-glucoside, ≥98% (HPLC), MilliporeSigma™ Supelco™
Description
Food and Beverages
Specifications
Specifications
| Packaging | Bottomless Glass Bottle with Inserted Fused Cone |
| CAS | 482-35-9 |
| Physical Form | Neat |
| Linear Formula | C21H20O12 |
| MDL Number | MFCD00017746 |
| Synonym | Quercetin 3-beta-D-glucoside; 3,3 ′,4 ′,5,7-Pentahydroxyflavone 3-beta-glucoside; Isoquercitrin |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 464.38 |
| Percent Purity | ≥98% (HPLC) |
| Show More |