missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Qubit™ RNA High Sensitivity (HS), Broad Range (BR), and Extended Range (XR) Assay Kits

Catalog No. p-7235447
Encompass
Change view
Click to view available options
Quantity:
100 assays
500 assays
Assay:
RNA Quantification, BR
RNA Quantification, ext range
RNA Quantification, HS
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Assay
Q10210 100 assays RNA Quantification, BR
Q10211 500 assays RNA Quantification, BR
Q33223 100 assays RNA Quantification, ext range
Q33224 500 assays RNA Quantification, ext range
Q32852 100 assays RNA Quantification, HS
Q32855 500 assays RNA Quantification, HS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. Q10210 Supplier Invitrogen™ Supplier No. Q10210
Only null left

Kits enable quick and sensitive detection of low and high abundance RNA, and can distinguish between RNA and DNA, protein, and free nucleotides.

Achieve accurate and precise quantification of RNA with Qubit RNA HS, BR, and XR Assay Kits. These RNA quantification kits enable quick and selective detection of low and high abundance RNA samples, and can distinguish RNA from DNA, protein, and free nucleotides. Contaminants such as salts, solvents, or detergents are well-tolerated.

Qubit RNA HS, BR, and XR Assay Kits, designed for use with Qubit Fluorometers, are highly selective for RNA over DNA, protein, and free nucleotides. All kits provide concentrated assay reagent, dilution buffer, and pre-diluted RNA standards. Simply dilute the reagent using the buffer provided, add your sample (any volume between 1 μL and 20 μL is acceptable), and read the concentration using a Qubit Fluorometer.

Qubit RNA HS Assay Kit
The Qubit RNA HS (High Sensitivity) Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of low-abundance RNA samples.  Depending on sample volume, the assay kit is designed to be accurate for initial RNA sample concentrations of 0.2 to 200 ng/μL, providing a detection range of 4−200 ng.

Qubit RNA BR Assay Kit
The Qubit RNA BR (Broad-Range) Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of RNA samples.  Depending on sample volume, the assay kit is designed to be accurate for initial RNA sample concentrations of 0.5 to 1,200 ng/μL, providing a detection range of 10−1,200 ng.

Qubit RNA XR Assay Kit

The Qubit RNA XR (Extended Range) Assay Kit, designed for use with Qubit 4 and Qubit Flex Fluorometers, provides an accurate and selective method for the quantitation of high-abundance RNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial RNA sample concentrations of 5 to 20,000 ng/μL, providing a detection range of 100−20,000 ng.

Notes
• Qubit RNA HS and BR Assay kits can be used with any Qubit Fluorometer
• Qubit RNA XR Assay Kits are designed to be used only with Qubit 4 or Qubit Flex Fluorometers
• Use with thin-wall, clear, 0.5-mL PCR tubes (Cat. No. Q32856) for the Qubit 4 Fluorometer and 8 x 200-μL tube strips (Cat. No. Q33252) for the Qubit Flex Fluorometer

Specifications

Assay RNA Quantification, BR
Detection Method Fluorescence
Excitation/Emission 644/673
For Use With (Equipment) Qubit Fluorometer
No. of Reactions 100 Reactions
Quantitation Range 10 to 1200 ng
Product Line Quant-iT
Quantity 100 assays
Shipping Condition Room Temperature