Learn More
Invitrogen™ Qubit™ RNA High Sensitivity (HS), Broad Range (BR), and Extended Range (XR) Assay Kits
Description
Achieve accurate and precise quantification of RNA with Qubit RNA HS, BR, and XR Assay Kits. These RNA quantification kits enable quick and selective detection of low and high abundance RNA samples, and can distinguish RNA from DNA, protein, and free nucleotides. Contaminants such as salts, solvents, or detergents are well-tolerated.
Qubit RNA HS Assay Kit
The Qubit RNA HS (High Sensitivity) Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of low-abundance RNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial RNA sample concentrations of 0.2 to 200 ng/μL, providing a detection range of 4−200 ng.
Qubit RNA BR Assay Kit
The Qubit RNA BR (Broad-Range) Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of RNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial RNA sample concentrations of 0.5 to 1,200 ng/μL, providing a detection range of 10−1,200 ng.
Qubit RNA XR Assay Kit
The Qubit RNA XR (Extended Range) Assay Kit, designed for use with Qubit 4 and Qubit Flex Fluorometers, provides an accurate and selective method for the quantitation of high-abundance RNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial RNA sample concentrations of 5 to 20,000 ng/μL, providing a detection range of 100−20,000 ng.
Notes
• Qubit RNA HS and BR Assay kits can be used with any Qubit Fluorometer
• Qubit RNA XR Assay Kits are designed to be used only with Qubit 4 or Qubit Flex Fluorometers
• Use with thin-wall, clear, 0.5-mL PCR tubes (Cat. No. Q32856) for the Qubit 4 Fluorometer and 8 x 200-μL tube strips (Cat. No. Q33252) for the Qubit Flex Fluorometer
Specifications
Specifications
| Assay | RNA Quantification, BR |
| Detection Method | Fluorescence |
| Excitation/Emission | 644/673 |
| For Use With (Equipment) | Qubit Fluorometer |
| No. of Reactions | 500 Reactions |
| Quantitation Range | 10 to 1200 ng |
| Product Line | Quant-iT |
| Quantity | 500 assays |
| Shipping Condition | Room Temperature |
Frequently Asked Questions (FAQs)
Here are several suggestions:
1.View the raw fluorescence value (RFU) for the standards under Check Standards or Check Calibration. Confirm that the values for the samples fall between the values of the standards (or a little above the highest standard). If they do not, the sample is out of the accurate range of the assay. Refer to the confidence ranges for each assay in the product manuals. The readout in the assay will be to 2 significant figures instead of 3 if the assay sample is out of the high confidence range.
To bring the sample into the accurate range, dilute the sample or use more or less of it (for example, 10 µL instead of 2 µL if the sample reads low).
2.Check for temperature issues: The assay is temperature sensitive and the fluorescent signal can decrease at higher temperatures. Temperature fluctuations between samples, or between samples and standards, can cause problems.
Make sure that the buffer and Qubit reagent in DMSO are at room temperature. The buffer and Qubit reagent should be stored at room temperature, not in the refrigerator. Even after 2-3 hours at room temperature, buffer previously stored at 4°C can remain below room temperature.
Make sure your samples and working solution are not too warm (including those straight from a centrifuge). Samples kept in the Qubit instrument too long or read multiple times can warm up. If you want to perform multiple readings of a single tube, you should remove the tube from the instrument and let it equilibrate to room temperature for 30 seconds before taking another reading. Also, do not hold tubes in your hand for very long before reading them in the instrument, since this can warm the sample, resulting in a low reading.
3.Ensure that you have prepared the Qubit working solution correctly (1:200 dilution using the buffer provided in the kit). Ensure that you have prepared the standard tubes correctly (10 µL of each standard in 190 µL of the working solution). Ensure that the tubes are filled with at least 200 µL (both standards and samples).
4.Ensure that the reagents and standards you are using are less than 6 months old, and that the standards have been stored correctly. The Qubit reagent stock solution should be protected from light as much as possible.
5.Ensure that you have selected the correct assay on the Qubit Fluorometer for the Qubit assay you are performing.
6.Ensure that the lid is completely closed when reading standards and samples.
7.Use recommended tubes (both so the tube does not obstruct the lid, and for optical clarity). Some types of tubes can have high autofluorescence that will affect the assay.
8.Did you enter the number of microliters of stock you pipetted into the working solution into the Qubit instrument? If so, the reading after giving the Qubit Fluorometer this information is the concentration of your stock solution. If you did not, the reading you got is for the concentration in the assay tube (the tube you put into the Qubit Fluorometer) and not your stock solution.
9.If you are comparing Qubit assay results to concentration obtained by UV absorbance, and the concentration based on absorbance is significantly higher, it may be because of nucleic acid or protein contamination. The Qubit assays are much more specific for DNA, RNA, or protein than absorbance readings.
Please see our suggestions below:
The exact excitation/emission wavelength information is proprietary. Here are the approximate excitation/emission wavelengths:
- Qubit dsDNA HS Assay: ~500 nm/ ~530 nm
- Qubit dsDNA BR Assay: ~510 nm/ ~530 nm
- Qubit ssDNA Assay: ~490 nm/ ~520 nm
- Qubit RNA HS Assay: ~640 nm/ ~670 nm
- Qubit RNA BR Assay: ~640 nm/ ~670 nm
- Qubit microRNA Assay: ~500 nm/ ~520 nm
- Qubit Protein Assay: ~470 nm/ ~570 nm
Yes, you can, for Qubit instruments developed after the original Qubit (1.0) Fluorometer. See MyQubit assay instructions here (http://www.thermofisher.com/us/en/home/life-science/laboratory-instruments/fluorometers/qubit/qubit-assays/myqubit.html.html).
Generally, the cleaner the sample the better. Some salts, proteins, and detergents are tolerated in the assays; see the specific assay protocol for which ones and at what concentrations.