missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Qubit™ Protein and Protein Broad Range (BR) Assay Kits

Catalog No. p-7235452
Encompass
Change view
Click to view available options
Assay:
Qubit Protein Assay
Qubit Protein Broad Range Assay
Quantitation Range:
0.1 to 20 mg/mL
12.5 μg/mL to 5 mg/mL
Sufficient For:
100 Reactions
500 Reactions
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Assay Quantitation Range Sufficient For
Q33211 Qubit Protein Assay 12.5 μg/mL to 5 mg/mL 100 Reactions
Q33212 Qubit Protein Assay 12.5 μg/mL to 5 mg/mL 500 Reactions
A50668 Qubit Protein Broad Range Assay 0.1 to 20 mg/mL 100 Reactions
A50669 Qubit Protein Broad Range Assay 0.1 to 20 mg/mL 500 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. Q33211 Supplier Invitrogen™ Supplier No. Q33211
Only null left

Requires

Qubit 1.0 or 2.0 Fluorometer or other fluorometer and 500μL thin-walled PCR tubes

Achieve sensitive detection of protein, and low protein-to-protein variation, with the Qubit Protein and Protein BR Assay Kits for fluorescence-based protein quantitation assays.

Quantify protein quickly and accurately with two unique Qubit protein quantification kits: the Qubit Protein Assay, which quantifies 12.5 μg/mL to 5 mg/mL of protein, and the Qubit Protein BR Assay Kit, which quantifies 0.1 to 20 mg/mL of protein. Both assays are compatible with many common contaminants, reducing reagents, salts, and nucleic acids. The Protein BR Assay Kit is also tolerant of detergents.

Qubit Protein Assay Kit
The Qubit Protein Assay Kit is designed specifically for use with any Qubit fluorometer. While using between 1 and 20 μL of sample, this assay can quantify samples ranging from 12.5 μg/mL to 5 mg/mL with low protein-to-protein variation.

The Qubit Protein Assay Kit is highly selective for proteins and is also highly accurate in the presence of reducing reagents, but not in the presence of large amounts of detergent. Common contaminants, such as reducing reagents (e.g., DTT, β-mercaptoethanol), salts, free nucleotides, amino acids, solvents, and DNA (but not detergents), are well tolerated in the assay. Slight protocol modifications are required for other contaminants.

The kit provides concentrated assay reagent, dilution buffer, and pre-diluted BSA standards. Simply dilute the reagent using the buffer provided, add your sample (any volume between 1 μL and 20 μL is acceptable), and read the concentration.

Which product should you choose for fluorometric protein quantitation?
• 1–20 samples: use this Qubit Protein Assay Kit with the Qubit Fluorometer
• 20–2000 samples: use the Quant-iT Protein Assay Kit with a microplate reader

Qubit Protein BR Assay Kit
The Qubit Protein BR Assay Kit provides a quick, easy, and reliable method for determining the protein concentration of purified proteins, lysates, serum, plasma, or complex protein mixtures using a Qubit 4 Fluorometer. This protein kit can deliver accurate protein quantitation across a broad range (0.1–20 mg/mL), enabling most samples to be used neat (undiluted) and eliminating the guess work and dilution steps that accompany traditional protein quantitation methods. The assay is compatible with many common contaminants, such as detergents, reducing reagents (e.g., DTT, β-mercaptoethanol), salts, and nucleic acids.

Key features of the Qubit Protein BR Assay Kit include:
Broad dynamic range—0.1 to 20 mg/mL, measure samples without dilution
Two-point standard curve—eliminate time-consuming standard preparation
Automatic concentration determination—receive protein concentration instantaneously; no need for offline calculations
Compatible—accurate in the presence of reducing reagents, detergents, and other common buffer components

The Qubit Protein BR Assay Kit comes with two calibration standards, Protein BR Assay Buffer, and Protein BR Assay Reagent. Simply add 150–160 μL of Protein BR Assay Buffer to 10 or 20 μL of your sample, followed by 30 μL of Protein BR Assay Reagent. Incubate for 10 minutes at room temperature. Then insert the sample into a Qubit 4 instrument and read the concentration.

Notes
• The Qubit Protein Assay Kit can be used with Qubit 1.0, Qubit 2.0, Qubit 3, and Qubit 4 fluorometers
• The Qubit Protein BR Assay Kit is a fluorescence-based assay intended for use only with the Qubit 4 Fluorometer
• 500 μL thin-walled clear PCR tubes (Cat. No. Q32856) are required but not included in the Protein Assay and BR Assay Kits

Order Info

Shipping Condition: Room temperature

Specifications

Assay Qubit Protein Assay
Detection Method Fluorescence
Product Line Qubit
Product Type Protein Quantification Assay
Sufficient For 100 Reactions
For Use With (Equipment) Qubit Fluorometer, Microplate Reader
Quantitation Range 12.5 μg/mL to 5 mg/mL
Quantity 100 Assays
Shipping Condition Room Temperature