missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Qubit NGS Library Quantification Assay Kit

Catalog No. Q34250
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q34250 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q34250 Supplier Invitrogen™ Supplier No. Q34250
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Invitrogen™ Qubit™ NGS Library Quantification Assay is an easy-to-use fluorescence-based assay for precise molar quantification of sequencing-ready Illumina next-generation sequencing (NGS) libraries using the Qubit™ Flex Fluorometer or compatible fluorescence microplate readers.

The Invitrogen™ Qubit™ NGS Library Quantification Assay is an easy-to-use fluorescence-based assay for precise molar quantification of sequencing-ready Illumina next-generation sequencing (NGS) libraries using the Qubit™ Flex Fluorometer or compatible fluorescence microplate readers. The assay selectively quantifies P5/P7-adapted double-stranded DNA libraries prior to sequencing, helping ensure optimal cluster density and successful sequencing runs.

Accurate determination of library concentration is a critical step in NGS workflows, as the amount of DNA loaded onto a flow cell directly impacts cluster density, data quality, and sequencing efficiency. The Qubit NGS Library Quantification Assay provides a dilution-free, time-saving alternative to qPCR-based library quantification methods while delivering comparable precision.

Key features and benefits:

  • Selective molar quantification—measures only dual-adapted, sequencing-ready libraries for improved accuracy
  • Dilution-free workflow—eliminates error-prone serial dilution steps required for qPCR
  • Broad dynamic range—quantifies libraries from 0.8–46 nM without additional dilution
  • Robust performance—accurately quantifies target libraries even in the presence of contaminating nucleic acids
  • Flexible detection—compatible with the Qubit Flex Fluorometer and fluorescence microplate readers
  • Accessible and cost-effective—no need for dedicated qPCR instrumentation

The streamlined workflow reduces hands-on time and minimizes potential user error, enabling efficient library quantification for both genomic core facilities and individual laboratories. By delivering precise quantification of sequencing-ready libraries, the Qubit NGS Library Quantification Assay helps ensure optimal cluster density on flow cells, improving sequencing outcomes.

Notes

  • Designed for use with the Qubit Flex Fluorometer (Cat. No. Q33327) and fluorescence-based microplate readers. The assay is not currently available for use on Qubit 4 Fluorometers.
  • Optimized for use with Qubit Flex Pyrogen Free Assay Tube Strips (Cat. No. Q32893).
  • Requires the use of the Qubit Flex System Verification Assay Kit (Cat. No. Q33254) prior to the first assay run.

Specifications

Detection Method Fluorescence
Format Kit
Isolation Technology Magnetic Bead
Sequencing Type Next Generation Sequencing
Excitation 490 nm
Emission 515 nm
No. of Reactions 96 Reactions
Product Type Fluorescence Assay
Quantity 1 Ea.

For Research Use Only. Not for use in diagnostic procedures.