missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Qubit and Quant-it Assay Kit Standards
GREENER_CHOICE

Catalog No. Q33233
Encompass
Change view
Click to view available options
Quantity:
1 set
5 each
5 mL
2 x 5 mL
Product Type:
1X dsDNA HS Assay Lambda Std
Qubit 1X dsDNA BR Lambda Std
Qubit RNA IQ Standards
Qubit RNA XR Standard
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
Q33233 5 mL 1X dsDNA HS Assay Lambda Std
Q33235 1 set Qubit RNA IQ Standards
Q33236 5 each Qubit RNA XR Standard
Q33263 2 x 5 mL Qubit 1X dsDNA BR Lambda Std
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. Q33233 Supplier Invitrogen™ Supplier No. Q33233
Save Up to $945 with a Cell Counter Starter Kit
View all promotions

Achieve precise quantitation of dsDNA or RNA samples with stand-alone Qubit nucleic acid standards.

Achieve precise quantitation of dsDNA or RNA samples with stand-alone Qubit nucleic acid standards. The Qubit standards can be used in conjunction with different Qubit or Quant-iT nucleic acid quantitation assay kits and offer stand-alone versions of the kit standard for performing additional assays.

Qubit 1X dsDNA HS Assay Lambda Standard

Qubit 1X dsDNA HS Assay Lambda Standard can be used with either Qubit 1X dsDNA HS Assay Kits, Qubit dsDNA HS Assay Kits, or their corresponding Quant-iT Assay Kits. The standards are supplied at the same concentration as the highest concentration dsDNA standard (10 ng/μL) in the Quant-iT Assay Kits and can be diluted into the standard series.

Qubit 1X dsDNA BR Standards

Qubit 1X dsDNA BR (Broad Range) Standards can be used with either Qubit 1X dsDNA BR Assay Kits, Qubit dsDNA BR Assay Kits, or their corresponding Quant-iT Assay Kits. The Qubit 1X dsDNA BR Standards includes standards at two concentrations, 0 and 100 ng/μL. The high standard corresponds to the same concentration as the highest concentration dsDNA standard in the Quant-iT Assay Kits and can be diluted into the standard series.

Qubit RNA IQ Assay Standards

Qubit RNA IQ Assay RNA standards are designed for use with the Qubit RNA IQ Assay Kit. Sufficient amounts of the small and large and/or structured RNA standard are supplied for generating an IQ standard curve for up to 500 samples. The RNA IQ standards are designed to measure RNA degradation and are not interchangeable with the standards from other RNA quantitation assays, including the Qubit RNA HS, BR, or XR assay kits.

Note: Use thin-wall, clear, 0.5-mL PCR tubes (Cat. No. Q32856) for the Qubit 4 fluorometer and 8 x 200-μL tube strips (Cat. No. Q33252) for the Qubit Flex fluorometer.

Specifications

Content And Storage Qubit™ dsDNA HS Assay – Lambda DNA Standard, 5 mL at 10 ng/μL in TE buffer
Store at 2°C to 8°C
Concentration 10 ng/μL
For Use With (Application) Qubit and Quant‐iT dsDNA HS Assays and 1 x dsDNA Assays
For Use With (Equipment) Qubit Fluorometer or fluorescence microplate reader
Green Features Fewer resources used, Less waste
Product Line Qubit
Product Type 1X dsDNA HS Assay Lambda Std
Quantity 5 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.