missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Qubit™ 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits
GREENER_CHOICE

Catalog No. Q33266
Encompass
Change view
Click to view available options
Quantity:
100 assays
500 assays
Quantitation Range:
0.1 to 120 ng
4 to 4000 ng
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Quantitation Range
Q33266 500 assays 4 to 4000 ng
Q33230 100 assays 0.1 to 120 ng
Q33231 500 assays 0.1 to 120 ng
Q33265 100 assays 4 to 4000 ng
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. Q33266 Supplier Invitrogen™ Supplier No. Q33266

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Highly selective for dsDNA over RNA, the Qubit 1X dsDNA HS and BR Assay Kits offer easy-to-use formulations for the accurate quantitation of dsDNA in 100 and 500 assay formats. Lambda Standards provide sufficient material for up to 500 samples.

Experience easy, accurate, and precise quantification of dsDNA with Qubit 1X dsDNA HS (High Sensitivity) and Qubit 1X dsDNA BR (Broad Range) Assay Kits. The 1X formulation provides a ready-to-use working solution that enables simple dsDNA sample quantification on Qubit Fluorometers. These assays are highly selective for dsDNA over ssDNA, RNA, protein, and free nucleotides. Contaminants, such as salts, solvents, or detergents are well-tolerated.

Qubit 1X dsDNA HS and BR Assay Kits, designed for use with Qubit Fluorometers, are highly selective for double-stranded DNA (dsDNA) over single-stranded DNA (ssDNA), RNA, protein, and free nucleotides. All kits provide a ready-to-use working solution and DNA standards. To perform the assay, simply dilute your sample (any volume from 1–20 μL is acceptable) into the 1X working solution provided, then read the concentration using a Qubit Fluorometer.

Qubit 1X dsDNA HS Assay Kit
The Qubit dsDNA HS (High Sensitivity) Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of sensitive DNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial DNA sample concentrations of 5 pg/μL to 120 ng/μL, providing a detection range of 0.1−120 ng.

Qubit 1X dsDNA BR Assay Kit
The Qubit dsDNA BR (Broad-Range) Assay Kit, designed for use with Qubit 4 or Qubit Flex Fluorometers, provides an accurate and selective method for the quantitation of DNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial DNA sample concentrations of 0.2 to 4,000 ng/μL, providing a detection range of 4-4,000 ng.

Notes
• Qubit 1X dsDNA HS can be used with Qubit 2, Qubit 3, Qubit 4, and Qubit Flex Fluorometers
• Qubit 1X dsDNA BR are designed for use with Qubit 4 and Qubit Flex Fluorometers
• Use thin-wall, clear, 0.5-mL PCR tubes (Cat. No. Q32856) for the Qubit 4 Fluorometer and 8 x 200 μL tube strips (Cat. No. Q33252) for the Qubit Flex Fluorometer

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay 1X dsDNA, broad range
Detection Method Fluorescence
For Use With (Equipment) Qubit 4 and Qubit Flex Fluorometer
No. of Reactions 500 Reactions
Quantitation Range 4 to 4000 ng
Product Line Qubit
Quantity 500 assays
Shipping Condition This assay kit will ship as two components.
• Qubit™ 1X dsDNA BR Working Solution will be shipped at room temperature.
• Qubit 1X dsDNA BR lambda standards will be shipped at room temperature.
What is the shelf life for the 1X dsDNA BR working solution(Cat. No. Q33262), that is part of the Qubit 1X dsDNA BR Assay (Cat Nos. Q33265, Q33266)?

The storage conditions for the 1X dsDNA BR working solution are 18–28 degrees C (room temperature). When stored as directed, the kit contents are stable for at least 6 months from delivery date. The standards should still be kept in the fridge.

After mixing the sample or the standard with the working solution, the fluorescence signal is stable for 3 hours at room temperature when samples are protected from light, following the incubation period.
Storage information can be found on page 1, and incubation time is on page 2, of the Qubit 1X dsDNA BR Assay User Guide

Does the Qubit Flex Quantitation Starter Kit (Cat. No. Q45894) include a Qubit dsDNA Broad Range Assay Kit (Cat. No. Q33265, Q33266)?

No. The Qubit Flex Quantitation Starter Kit (Cat. No. Q45894) does not include a Qubit dsDNA Broad Range Assay Kit (Cat. No. Q33265, Q33266). It must be purchased separately.

The description of the Qubit Flex Starter Packages (Cat. Nos. Q32894, Q45893, Q45894) can be found on the Qubit Flex Starter Packages product page.

What is the expiration date for Qubit 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits (Cat. Nos. Cat. Q33230, Q33231, Q33265, Q33266)?

The Qubit 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits (Cat. Nos. Cat. Q33230, Q33231, Q33265, Q33266) are stable for 6 months from the date of receipt when stored as directed.

For more information on product storage, please refer to the user guides:

Qubit 1X dsDNA HS Assay Kit and Qubit 1X dsDNA BR Assay Kit

How should I store my Qubit 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits (Cat. No. Q33230, Q33231, Q33265, Q33266)?

The Qubit 1X dsDNA High Sensitivity (HS) (Cat. Nos. Q33230, Q33231) and Broad Range (BR) Assay Kits (Cat. Nos. Q33265, Q33266) have different storage conditions.

For Qubit 1X dsDNA HS Assay Kits (Cat. Nos. Q33230, Q33231), store all components at 2 - 8 degrees C.

For Qubit 1X dsDNA BR Assay Kits (Cat. Nos. Q33265, Q33266), store the working solution at 18 - 28 degrees C. Store the standards at 2 - 8 degrees C.

The assay kits are stable for at least 6 months from the date of receipt.

For more information, please refer to these user guides: Qubit 1X dsDNA HS Assay Kit and Qubit 1X dsDNA BR Assay Kit