Learn More
Bel-Art™ Quaver™ and Quaverette™ Vibrating Spatulas
Accurately transfer small amounts of material
$270.56 - $389.24
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) Weighing dish or weighing boat or container
Material Stainless Steel
View More Specs
Products 3
Catalog Number Mfr. No. Product Type Blade Material Shape Length (English) Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Blade Material Shape Length (English) Price Quantity & Availability  
22385039
View Documents Promotion Details
Bel-Art™
H367140000
Quaver™ Vibrating Spatula Stainless Steel Flat, Curved End 7 in.
Each for $270.56
Case of 24 Each for $6,493.44
Only null left
14430
View Documents Promotion Details
Bel-Art™
H367180000
Quaverette™ Micro-Blade Vibrating Spatula Stainless Steel Narrow 8.25 in.
Each for $272.55
Case of 24 Each for $6,541.20
Only null left
03422601
View Documents Promotion Details
Bel-Art™
H367190000
Quaverette™ Micro-Blade Vibrating Spatula Stainless Steel, Fluo-Kem™ Teflon™ FEP Coating Narrow 8.25 in.
Each for $389.24
Case of 24 Each for $9,341.76
Only null left
Description
Description
  • Rotating the toothed thumb wheel vibrates the spatula, giving user control over transfer from spatula to weighing dish
  • Make accurate additions of precious materials as needed
  • Quaver™ spatula features a flat blade with curved end
Specifications
Specifications
Not Autoclavable
Weighing dish or weighing boat or container
Stainless Steel