Learn More
Applied Biosystems™ QuantStudio™ 7 Flex Real-Time PCR System, array card, desktop
Description
Includes
Desktop Computer
With a simplified workflow, intuitive software, automation capabilities, and touch-screen interface, the QuantStudio™ 7 Flex system offers exceptional reproducibility with minimal well-to-well and instrument-to-instrument variation.
- Increased application versatility through the accommodation of 4 different block types
- Compatible with over 8 million TaqMan™ assays for gene expression, genetic variation, gene regulation, or protein expression experiments
- The intelligent design of the QuantStudio™ platform combined with Applied Biosystems™ instrument reliability and accuracy
- Easy-to-use software, responsive touch-screen, automation capabilities, and effortless block exchange without the need for any tools
Thermo Fisher Cloud:
Thermo Fisher Cloud provides scientists with a secure place to store, analyze, and share data. Applied Biosystems qPCR Analysis Modules within the Thermo Fisher Cloud are web browser-based software tools that are flexible, fast and easy to use. These software tools are compatible with Mac or PC as well as most Applied Biosystems instrument file types.
The QuantStudio™ 7 Flex system accommodates the interchange of a 96-well, 96-well Fast, 384-well, or TaqMan™ array card block. The system is delivered with one block type (an array card block in this case); other block types may be purchased separately. In addition, a Twister™ Robot Automation Accessory is available for this system to support high throughput workflows. For a more value-priced system that does not support TaqMan™ array cards or automation (but that can be upgraded to do so at a later date), please see our QuantStudio™ 6 Flex system. And for the ultimate in flexibility, consider the QuantStudio™ 12K Flex system, which also accommodates TaqMan™ OpenArray™ plates.
Accomplish More, Faster:
Run hundreds of real-time PCR reactions effortlessly using TaqMan™ Array 384-well microfluidic cards. And when integrated with the Twister™ Robot Automation Accessory (available separately), the QuantStudio™ 7 Flex system maximizes your throughput capabilities.
High Application Versatility:
Simple workflows, reliable assays and reagents, multiple formats, and intuitive software are available for a broad range of applications, including gene expression, genetic variation, gene regulation, High Resolution Melt (HRM), and Protein Thermal Shift™ experiments.
Results You Can Trust:
Improved well-to-well and instrument-to-instrument data accuracy is achieved with the OptiFlex™ system, which features 6 decoupled excitation and emission filter channels with 21 filter combinations for maximum multiplexing and chemistry flexibility.
Sensitive Data Analysis:
Detect changes in target quantity as small as 1.5-fold in single-plex reactions and obtain 10 logs of linear dynamic range.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
| Block Format | Interchangeable |
| Format | 384-well Microfluidic Card |
| Content And Storage | Laptop Computer |
| For Use With (Equipment) | QuantStudio™ |
| Product Line | QuantStudio |
| Display Type | Touchscreen |
| Quantity | 1 system |
Frequently Asked Questions (FAQs)
Please follow the instructions found in the Software Licensing Quick Reference Guide at https://fnoclient.gss.tf/quick-start-guide to activate or renew a license for the QuantStudio 6 or QuantStudio 7 Real-Time PCR System. If you have a license key associated with a computer that is no longer in use, please follow the instructions to "Transfer a license to a different user or computer" in the above guide.
After launching the QuantStudio software, go to the Maintenance Manager. Check if the QuantStudio system icon is present in the On the Network window. If not, click on the Refresh' button to the left of Remove from My Instruments'. If you still do not see the QuantStudio system icon for the following reasons:
- There is an icon but you cannot add it into the "My Instruments" area
- There is a red X on the instrument
- A message says "Disconnected" at the bottom of the window with a red X in it
then you will need to add the Instrument into the My Instruments' area. Click on the wrench icon in the upper right-hand corner of the icon as pictured below. You may get a message that says the firmware is older than the one you're trying to run. If appropriate, choose to download the new firmware and let it progress to finish. Do not turn off the instrument during this update. After the firmware update is completed, restart the instrument and check for the QuantStudio system icon in On the Network' area.
This error can occur due to allocation of the computer's virtual memory. If you see this message, try the following:
1. Click OK' to the error, close any unnecessary programs, and re-launch the software (you may have to do this several times).
2. Using the Task Manager (Control+Alt+Delete), look for a process called javaw.exe'. Select this line and end the process. Then re-launch the software.
The calibration files are stored on the QuantStudio 6/7 Flex instrument itself; therefore, when you need to perform a new calibration, you will have to delete the old calibration files. From the instrument console, go to the QuantStudio 6/7 Flex instrument touchscreen and select Collect Results'. You should then see a list of files stored on the instrument. Manually select a calibration file such as Fast-96-cal, and press the Delete' button to remove the old calibration file. Occasionally, users cannot delete the file off the instrument and may encounter this error: Some experiments did not collect results therefore cannot be deleted.. In these situations, launch the software, go to maintenance', manage files, and download the calibration file. The status of the experiment should be displayed as complete'. Next, delete the old calibration file and rerun the calibration.
The reveal traces' feature for genotyping runs allows you to trace the clusters throughout the PCR process. For some assays you may find better cluster discrimination at, say, cycle 40, as opposed to cycle 50. To use this feature, the amplification data needs to have been collected for the run.
- Go to Analysis Settings'
- Under the default Call Settings' tab, Choose Analyze Real-Time Rn Data'
- Click Apply Analysis Settings'
- Under the Allelic Discrimination Plot, check the box next to Reveal Traces'. You can then move the slide bar to see how the data changes with the cycles number.
In the example below, the image on the left shows the data at a full 40 cycles. The image on the right is the same data after revealing traces (grey lines). The clusters can be traced back by moving the cycle bar. Notice that one point is no longer called at the earlier cycle set point.
For Research Use Only. Not for use in diagnostic procedures.