missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio™ 7 Pro Dx Real-Time PCR System, 96-well, 0.2 mL, desktop

Catalog No. A51192
Change view
Click to view available options
Quantity:
1 system
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A51192 1 system
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A51192 Supplier Applied Biosystems™ Supplier No. A51192

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

QuantStudio™ 7 Pro Dx, an In Vitro Diagnostic (IVD) Real-Time PCR system. Combines modern hardware and intuitive software to achieve maximum efficiency, smarter productivity, and higher accuracy for molecular diagnostic workflows.

The Applied Biosystems QuantStudio 7 Pro Dx Real-Time PCR System adds innovative smart features to the real-time PCR (qPCR) workflow, all in a compact footprint. With an open IVD format, voice commands, Smart Help, multiplex capability, simplified workflow, intuitive software, automation compatibility, and large capacitive touch-screen interface, the QuantStudio 7 Pro Dx system offers exceptional reproducibility with minimal well-to-well and instrument-to-instrument variation. Designed for ultimate ease-of-use, the system provides high quality, excellent reliability, and an optimal user experience for clinicians and assay developers who want to work smarter in the lab.

The system is US-IVD and IVDR compliant.

Flexibility to develop or diagnose
Flexible combined-use software on a single system simplifies the process from development through IVD.

Effortless workflow
Reduce manual work and user errors through an on-device, assay-driven workflow. Preset assay definition consists of run method, analysis, and report settings.

Compact stand-alone system
Set up a run, lay out assays, control the instrument, conduct plate analysis, send data to LIS, and generate a report directly from the touchscreen easily without a computer.

Adaptable sample throughput
The system offers added flexibility to accommodate different sample throughputs with interchangeable 96-well (0.2 mL) and 384-well blocks.

Peace of mind with Smart service
Minimize downtime with premium service offerings for the QuantStudio 7 Pro Dx system. Smart Help allows users to contact Technical Support directly from the touchscreen, without the need to download files or spend time on the phone. Experienced technicians will analyze the request and respond according to the user’s preference (phone or email) and resolve the issue as quickly as possible.

Smart Remote Support is an immersive collaboration tool that allows customers to quickly and efficiently show issues to Technical Support for resolution. Reduce instrument downtime compared to on-site repair and help minimize the need for on-site visits.

Easy and convenient hands-free operation
The QuantStudio 7 Pro Dx system was designed for a simple and powerful qPCR experience. Hands-free operation with voice commands enables you to quickly and easily start a run without even removing your gloves.

Connectivity for optimal productivity
Access, analyze, and share data anytime, anywhere with our Connect cloud-based platform and Instrument Connect mobile app. Monitor your run, securely store data, and share results online.

Results you can trust
Well-to-well and instrument-to-instrument data accuracy is achieved with the OptiFlex system, which features six decoupled excitation and emission filter channels with 21 filter combinations for maximum multiplexing and chemistry flexibility.

Sensitive data analysis
Detect changes in target quantity as small as 1.5-fold in single-plex reactions and obtain 10 logs of linear dynamic range.

Qualification services help enable regulatory compliance
Installation, operational, and performance qualification (IQOQPQ) services enable compliance with regulatory requirements. IQOQ is included with installation, and OQPQ is recommended after moving the instrument, upgrading software or hardware, planned maintenance, or critical repairs.

Get up and running fast with SmartStart Orientation
SmartStart Orientation is performed by a Field Application Scientist (FAS) remotely or in your lab and helps you:
• Get up and running quickly
• Get connected to your local network and cloud
• Choose appropriate reagents and consumables to meet your needs

Compliance

For In Vitro Diagnostic Use

Specifications

Block Format Interchangeable
Format 96-well
Content And Storage • 1 QuantStudio 7 Pro Dx Real Time PCR System
• 1 96-well, 0.2 mL block
• 1 96-well, 0.2 mL heated cover
• 1 Dell tower computer with monitor and keyboard
• 1 QuantStudio 7 Pro Dx IVD software kit
• 1 QuantStudio 7 Pro Dx IVD user documentation kit
• 1 Installation and qualification service

Dimensions 54.7 x 33.8 x 52.5 cm (H x W x D)
Product Line QuantStudio
Display Type 12 in. Capacitive Touchscreen
For Use With (Application) Infectious diseases, Oncology, Diagnostics
Shipping Condition Room Temperature
Communications Thermo Fisher Connect Platform, USB, LAN, or WiFi
Quantity 1 system
Weight 38 kg
Show More Show Less
What is the difference between continuous melt curves and step and hold melt curves on the Applied Biosystems Real-Time PCR Instruments?

Continuous melt curve: increases the temperature by the ramp increment (degrees C/sec) so that the temperature will change at a constant rate set in the protocol.
Step and Hold melt curve: increases the temperature by the ramp increment (degrees C) and then holds at that temperature for the time specified by the user.

The amount of data collected will depend on the filter(s) selected. The more filters you use, the longer the acquisition will take and the fewer data points will be collected.

I recently switched the block in my real-time PCR instrument. Now it shows the status as 'not calibrated' for all the calibration plates, even though my calibrations are current. What is causing this issue?

The issue might be due to the block not being properly seated in the instrument. Please follow the steps below:

  1. Remove the block from the instrument.
  2. Reinsert the block into the instrument.


If the problem persists after reseating the block, please contact our Technical Support team for further assistance.

For In Vitro Diagnostic Use.