Learn More
Applied Biosystems™ QuantStudio™ 6 Flex Real-Time PCR System, 384-well, desktop
Description
Includes
Desktop computer
With a simplified workflow, intuitive software, automation capabilities, and touch-screen interface, the QuantStudio™ 7 Flex system offers exceptional reproducibility with minimal well-to-well and instrument-to-instrument variation.
Features of the QuantStudio™ 6 Flex Real-Time PCR System include:
- Affordable price allows you to obtain a high performance instrument with minimal upfront capital investment
- Compatible with over 8 million TaqMan™ assays for gene expression, genetic variation, gene regulation, or protein expression experiments
- The intelligent design of the QuantStudio™ platform combined with Applied Biosystems™ instrument reliability and accuracy
- Easy-to-use software, responsive touch-screen, and effortless block exchange without the need for any tools
Thermo Fisher Cloud:
Thermo Fisher Cloud provides scientists with a secure place to store, analyze, and share data. Applied Biosystems qPCR Analysis Modules within the Thermo Fisher Cloud are web browser-based software tools that are flexible, fast and easy to use. These software tools are compatible with Mac or PC as well as most Applied Biosystems instrument file types.
The QuantStudio™ 6 Flex system accommodates the interchange of a 96-well, 96-well Fast, or 384-well block. The system is delivered with one block type (a 384-well block in this case); other block types may be purchased separately. For a system that also supports TaqMan™ array cards, as well as automation, please see our QuantStudio™ 7 Flex system. The QuantStudio™ 6 Flex system can also be upgraded to a QuantStudio™ 7 Flex system at a later date, as your needs change. For the ultimate in productivity, consider the QuantStudio™ 12K Flex system, which also accommodates TaqMan™ OpenArray™ plates.
Upgrade Capabilities When You Need Them:
The QuantStudio™ 6 Flex system can be seamlessly upgraded to a QuantStudio™ 7 Flex system by a field service engineer to give you additional automation, throughput, and multiplexing capabilities.
Results You Can Trust:
Improved well-to-well and instrument-to-instrument data accuracy is achieved with the OptiFlex™ system, which features 5 coupled excitation and emission filter channels.
Sensitive Data Analysis:
Detect changes in target quantity as small as 1.5-fold in single-plex reactions and obtain 10 logs of linear dynamic range.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
| Block Format | Interchangeable |
| Format | 384-well Plate |
| Content And Storage | Desktop Computer |
| For Use With (Equipment) | QuantStudio™ |
| Product Line | QuantStudio |
| Display Type | Touchscreen |
| Quantity | 1 system |
Frequently Asked Questions (FAQs)
We recommend that you send the run file to the QuantStudio 6 or QuantStudio 7 Real-Time PCR instrument and start the run from the instrument touchscreen. Additionally, you can upgrade to v1.3 or above of the QuantStudio Real-Time PCR software and you will not have this error message show up.
Please follow the instructions found in the Software Licensing Quick Reference Guide at https://fnoclient.gss.tf/quick-start-guide to activate or renew a license for the QuantStudio 6 or QuantStudio 7 Real-Time PCR System. If you have a license key associated with a computer that is no longer in use, please follow the instructions to "Transfer a license to a different user or computer" in the above guide.
After launching the QuantStudio software, go to the Maintenance Manager. Check if the QuantStudio system icon is present in the On the Network window. If not, click on the Refresh' button to the left of Remove from My Instruments'. If you still do not see the QuantStudio system icon for the following reasons:
- There is an icon but you cannot add it into the "My Instruments" area
- There is a red X on the instrument
- A message says "Disconnected" at the bottom of the window with a red X in it
then you will need to add the Instrument into the My Instruments' area. Click on the wrench icon in the upper right-hand corner of the icon as pictured below. You may get a message that says the firmware is older than the one you're trying to run. If appropriate, choose to download the new firmware and let it progress to finish. Do not turn off the instrument during this update. After the firmware update is completed, restart the instrument and check for the QuantStudio system icon in On the Network' area.
This error can occur due to allocation of the computer's virtual memory. If you see this message, try the following:
1. Click OK' to the error, close any unnecessary programs, and re-launch the software (you may have to do this several times).
2. Using the Task Manager (Control+Alt+Delete), look for a process called javaw.exe'. Select this line and end the process. Then re-launch the software.
The calibration files are stored on the QuantStudio 6/7 Flex instrument itself; therefore, when you need to perform a new calibration, you will have to delete the old calibration files. From the instrument console, go to the QuantStudio 6/7 Flex instrument touchscreen and select Collect Results'. You should then see a list of files stored on the instrument. Manually select a calibration file such as Fast-96-cal, and press the Delete' button to remove the old calibration file. Occasionally, users cannot delete the file off the instrument and may encounter this error: Some experiments did not collect results therefore cannot be deleted.. In these situations, launch the software, go to maintenance', manage files, and download the calibration file. The status of the experiment should be displayed as complete'. Next, delete the old calibration file and rerun the calibration.
For Research Use Only. Not for use in diagnostic procedures.