missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QuantStudio™ 3D Local Appliance/Server
Specifications
Specifications
| Content And Storage | • Dell PowerEdge T130 Tower • 23in Monitor • Keyboard • Mouse • 2 USB drives with software |
| Product Line | QuantStudio |
| Shipping Condition | Room Temperature |
| Quantity | 1 appliance/server |
For Research Use Only. Not for use in diagnostic procedures.