missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ QuantStudio™ 3D Digital PCR Tilt Base For GeneAmp™ PCR System 9700
Description
Includes
Instrument Base
Simply place the Tilt Base beneath the feet of the thermal cycler, load your prepared 3D digital PCR chips, and begin PCR amplification.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
| Content And Storage | Instrument Base |
| For Use With (Application) | Digital PCR, Real Time PCR (qPCR) |
| Quantity | 1 Base |
| Block Configurations | 3D Digital PCR Chips |