missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio 3 0.2 mL and SYBR Master Mix Starter Package

Catalog No. A39941
Change view
Click to view available options
Capacity:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
A39941 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A39941 Supplier Applied Biosystems™ Supplier No. A39941*

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Complete qPCR workflow bundle featuring the QuantStudio 3 0.2 mL system with laptop, SuperScript IV VILO Master Mix with ezDNase, PowerUp SYBR Master Mix, MicroAmp 96-well plates, optical adhesive covers, and SmartStart setup for fast, reliable gene expression analysis.

Enable efficient and reliable real-time PCR with this comprehensive, ready-to-use workflow solution. The QuantStudio™ 3 0.2 mL qPCR System bundle combines advanced instrumentation, optimized reagents, and essential consumables—along with a dedicated laptop and SmartStart onboarding support—to streamline setup and accelerate results from day one.

Designed for research laboratories focused on gene expression analysis and routine qPCR applications, this all-in-one package simplifies procurement while ensuring compatibility across every step of the workflow.

What's Included

  • QuantStudio™ 3 0.2 mL Real-Time PCR System
  • Pre-configured laptop for instrument control and data analysis
  • SmartStart installation and onboarding support for QuantStudio™ 3
  • SuperScript™ IV VILO™ Master Mix with ezDNase™
  • PowerUp™ SYBR™ Green Master Mix (5 x 5 mL)
  • MicroAmp™ 96-well reaction plates
  • MicroAmp™ optical adhesive covers

Features

  • Complete qPCR workflow solution: Includes instrument, software-ready laptop, reagents, and consumables in a single, convenient bundle to reduce sourcing complexity.
  • Flexible 0.2 mL block format: Supports standard reaction volumes and compatibility with widely used plate formats for versatile lab workflows.
  • High-performance real-time PCR system: The QuantStudio™ 3 system delivers precise thermal cycling, sensitive fluorescence detection, and intuitive software for consistent, high-quality results.
  • Rapid setup with SmartStart support: Guided installation and onboarding help users quickly implement the system and begin generating data with confidence.
  • Optimized reverse transcription: SuperScript™ IV VILO™ Master Mix with ezDNase™ enables efficient cDNA synthesis while helping reduce genomic DNA contamination.
  • Reliable SYBR Green chemistry: PowerUp™ SYBR™ Green Master Mix supports robust amplification performance, high specificity, and reproducibility across experiments.
  • Efficient plate-based workflow: MicroAmp™ 96-well plates support streamlined sample processing and consistent thermal performance.
  • Secure sealing for data integrity: Optical adhesive covers minimize evaporation and ensure reliable fluorescence detection.

Benefits

  • Accelerate time to results with a fully integrated system that reduces setup time and enables immediate workflow implementation.
  • Simplify procurement by consolidating instrumentation, reagents, and consumables into a single SKU.
  • Ensure workflow compatibility with components designed and validated to work seamlessly together.
  • Enhance data reliability through precise instrumentation and optimized reagent performance.
  • Support reproducibility across gene expression and routine qPCR applications.
  • Improve lab efficiency with a complete solution that reduces coordination across multiple suppliers.

This all-in-one QuantStudio™ 3 0.2 mL qPCR workflow bundle is designed to help laboratories streamline operations, reduce implementation time, and achieve consistent, high-quality real-time PCR results.

Specifications

Block Format Non-interchangeable
Capacity 1 x 96-well Plate
Voltage 100/240 V
Product Type Real Time PCR Instrument
No. of Programs 1000
Block Configurations 96-well