missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio™ 12K Flex Real-Time PCR System, Fast 96-well block, desktop

Catalog No. 4471088
Encompass
Change view
Click to view available options
Quantity:
1 system
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4471088 1 system
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4471088 Supplier Applied Biosystems™ Supplier No. 4471088

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Desktop Computer

QuantStudio™ 12K Flex system is a highly flexible, comprehensive RT-PCR platform that leverages the best of Applied Biosystems™ real-time technology in a single instrument.

The QuantStudio™ 12K Flex Real-Time PCR System includes easy-to-change blocks that can accommodate OpenArray™ plates, TaqMan™ array cards, 384-well, 96-well, and FAST 96-well plates allowing you to select the format that is right for your project whether it’s now or in the future, for maximum flexibility and consistency in your research.

The QuantStudio™ 12K Flex system is a highly flexible, comprehensive real-time PCR platform that leverages the best of Applied Biosystems real-time technology in a single instrument. The QuantStudio™ 12K Flex Real-Time PCR System includes easy-to-change blocks that can accommodate OpenArray™ plates, TaqMan™ array cards, 384-well, 96-well, and FAST 96-well plates allowing you to select the format that is right for your project whether it’s now or in the future, for maximum flexibility and consistency in your research.

Thermo Fisher Cloud
Thermo Fisher Cloud provides scientists with a secure place to store, analyze, and share data. Applied Biosystems qPCR Analysis Modules within the Thermo Fisher Cloud are web browser-based software tools that are flexible, fast and easy to use. These software tools are compatible with Mac or PC as well as most Applied Biosystems instrument file types.

The QuantStudio™ 12K Flex System’s intuitive software, easy touch screen setup, automation integration, effortless block exchange, and easy-to- use OpenArray™ components are designed to let you get started right away and reduce hands on time. Plan for the future and save time, money, and space in your lab with the only all-in-one qPCR instrument enabling a broad range of applications with maximum throughput.

Flexibility and Scalability
Includes five interchangeable blocks: The QuantStudio™ 12K Flex blocks accomodate 96-well, FAST 96-well, 384-well, TaqMan™ array cards, and OpenArray™ plates, for effortless scaling from 1 to 12,000 data points in a single run. Increase throughput as you dive deeper into your research, using just one instrument. As you begin your project, start with 96-well format experiments and then scale up to a higher throughput to get the statistically significant answers you need. Additionally, you can easily validate and screen your targets of interest with a selected panel of genes using novel formats such as the TaqMan™ array cards and OpenArray™ plates.
• Compatible with a full range of TaqMan™ assays:
The QuantStudio™ 12K Flex System is optimized to enable clear, clean data for all TaqMan™ assays that might be used across a broad range of applications, including gene expression, microRNA and small noncoding RNAs, SNP genotyping, protein analysis, protein thermal shift, high resolution melt, copy number variation, and pathogen detection.
• Provides a seamless switch from qPCR to digital PCR:
Increase the precision and sensitivity of your nucleic acid experiment by switching from qPCR to digital PCR mode using QuantStudio™ Digital PCR kits and DigitalSuite™ Software with the QuantStudio™ OpenArray™ block.
• Delivers peace of mind:
Leave early on a Friday and monitor runs from home, knowing that the system can safely store over 100 runs.

Maximum Throughput
Enhanced OptiFlex™ System: Building upon Applied Biosystems’ proprietary technology, the Enhanced OptiFlex™ System provides enhanced fluorescence detection for the accurate and sensitive data analysis necessary to collect 12,000 data points in a single run.
• Unprecedented coverage and sample throughput:
The OpenArray™ format accelerates genomic validation and screening, driving over 150,000 data points per work day, enabling most projects to be completed in days instead of weeks.
• Simple OpenArray™ workflow:
QuantStudio™ Open Array™ plates arrive in a case to facilitate ease of handling and ensure optimal data quality upon loading of samples using the Accufill™ System. Easily load four arrays into the QuantStudio™ 12K Flex instrument to achieve over 12,000 data points. Integrated sample tracking software lets you easily track your samples throughout the course of your project.
• Integrated analysis and quality systems:
New intuitive touch-screen interface and integrated calendar feature allows for scheduling time of use on the instrument. Additional analysis software such as DigitalSuite™ v1.0, ExpressionSuite v1.0, and Genotyper v1.1 is available to help you analyze hundreds of targets and samples and achieve the statistical power you need.

QuantStudio™ 12K Flex Software
• Intuitive interface and innovative design for novice and experienced users
• Convenient walk-away operation
• Improved data analysis across block types
• Fully compatible with high-throughput environments

Expandable Software Architecture
Optional add-ons are available to upgrade the existing software:
21 CFR Part 11 compliance: This optional SAE module assists with Security, Auditing, and Electronic signature of records for full data traceability.
• High Resolution Melt (HRM):
Using the built-in MeltDoctor™ protocols and calibrations, you can spend less time optimizing the run and more time customizing your results. Define the number of variants you want to see by quickly changing analysis settings.
• DigitalSuite™ Software v1.0:
For OpenArray™ block users needing sensitivity for rare targets and precision to measure small differences in target abundance.
• ExpressionSuite Software v1.0:
our latest stand-alone gene expression software with enhanced analysis tools that can analyze over 200 384-well plates or 10 OpenArray™ plates.

Specifications

Block Format Interchangeable
Format 96-well Plate
Sensitivity Down to 1 Copy
Reaction Volume Range 10–30 μL (96-well Fast Plate)
Dynamic Range Up to 9 Logs of Linear Dynamic Range
Content And Storage This instrument comes with an Fast 96-well Card Block.
High-throughput Compatibility High-throughput Compatible, Multiplexing
For Use With (Equipment) QuantStudio™
Reaction Speed Fast
Detection Method SYBR, Primer-Probe Detection
Optics 6 Emission Filters (500-720 nm), 6 Excitation Filters (450-670 nm), 21 Color Options
Dimensions 51 cm (W) x 74 cm (H) x 67 cm (D)
External Computer Computer and Monitor
Voltage 100 to 240 V
Electrical Requirements 60 Hz
Product Line QuantStudio
Display Type Touchscreen
Other Format Options 96-well Standard Block Upgrade Option, OpenArray™ Block Upgrade Option, TaqMan™ Array Card Block Upgrade Option, 384-well Block Upgrade Option
Programs Remote Monitoring of up to 10 Instruments
Quantity 1 system
Weight 69 kg (152 lb.)
Show More Show Less
How do I activate or renew my Vector NTI software license?

Please follow the instructions listed in this manual (https://licensing.appliedbiosystems.com/web/static/tools/manual.pdf).

For problems with license renewal, please send an email to instrumenthardwaresupport@thermofisher.com (for North America customers) or eurotech@thermofisher.com (for EMEA customers), with the computer ID and license key (AID-number) so you can be assisted.

When using the QuantStudio 12K Flex Real-Time PCR System, why are there no CRT values upon export of my data?

When this happens, the analysis was set to use ‘Relative Threshold' but the CRT values do not show up in the exported data. (Note: To use ‘Relative Threshold', go to “Analysis Settings”, and under the default Ct Settings tab, choose ‘Relative Threshold' under “Algorithm Settings”.)

To export the data, go to ‘Export' and make sure the ‘Results' section is checked. You will notice that the column header will still say ‘Ct' even if you have CRT in the well table like above. If you export the data but notice that the numbers still correspond to the ‘Ct' values (and not the CRT values), then you will need to use the ‘Load Export Set' button. Just click ‘OK' to the pop-up window. and the changes will be applied. The next time you export, the values should reflect the CRT values.

What can I do if I get the Error - "Interrupted data file" in the QuantStudio12K Flex software?

Sometimes an error can occur when the file is downloaded to the computer. When this occurs you may see an abrupt break in the data. If you see this, you can repair the data by going to Analysis then Analyze from Raw Data. You should then see a fixed plot. Please note that you would need to have at least v1.2 of the QuantStudio 12K Flex software to use this feature. If you do not have this version, or if you do but the error persists, please send your data file to techsupport@thermofisher.com so that we can attempt to recover the data.

Is it possible to see the traces in an allelic discrimination experiment in the QuantStudio 12K Flex Real-Time PCR System software?

The ‘reveal traces' feature for genotyping runs allows you to trace the clusters throughout the PCR process. For some assays you may find better cluster discrimination at, say, cycle 40, as opposed to cycle 50. To use this feature, the amplification data needs to have been collected for the run.

- Go to 'Analysis Settings'.
- Under the default ‘Call Settings' tab, Choose 'Analyze Real-Time Run (?) Data'.
- Click ‘Apply Analysis Settings'.
- Under the Allelic Discrimination Plot, check the box next to ‘Reveal Traces'. You can then move the slide bar to see how the data change with the cycle number.

How do I transfer a QuantStudio 12K Flex software license to another computer?

Please follow the directions in this user guide (https://licensing.appliedbiosystems.com/web/static/tools/manual.pdf) to manage your license (including transfer and renewing a license).

If you are unable to resolve the issue after following these steps, please contact us at instrumentservices@thermofisher.com for further assistance.

Where are my files saved in the QuantStudio 12K Flex Real-Time PCR System software?

Run files will be saved to both the instrument and the connected computer. On the connected computer, files will be saved to the default data folder, unless you change it. To find or change the default folder, go to Tools then Preferences then Defaults. Here you will see a Data Folder and an Import Folder. The default location is shown. If you want files to be saved to (or open from) a different location, click ‘Browse' and choose the new folder.

What volumes can be used in my plate on the QuantStudio 12K Flex Real-Time PCR System?

The following volumes are supported for each instrument block:

- 96-well standard (10-100 µL reactions)
- 96-well Fast (10-30 µL reactions)
- 384-well (5-20 µL reactions)
- TaqMan Array Micro Fluidic Cards (approximately 1 µL reactions)
- OpenArray Plate (33 nL)

How many files can the QuantStudio 12K Flex Real-Time PCR System hold?

The QuantStudio 12K Flex Real-Time PCR System can store up to 100 runs on the instrument itself.

How do I add a custom dye to the QuantStudio 12K Flex Real-Time PCR System?

You will need an oligo sequence with the custom dye but without the quencher molecule. Set up a plate of dye dilutions such as seen in the “Custom Dye Calibration” section of this link:
https://tools.thermofisher.com/content/sfs/manuals/4474347B.pdf
1.Set up a dummy run using the ‘Standard Curve' option. Alter the thermal profile so that it simply ramps to 60 degrees C with a 2 min hold. Ensure that the filters of interest are selected.
2.When the run is complete, export and examine the raw data. Select the concentration to use by finding the dilution that will give you an acceptable signal in the following ranges:
- For a 384-well plate: between 400,000 and 1,200,000
- For a 96-well plate: between 1,400,000 and 4,300,000
3.Create a full plate of the dye using the selected concentration and run the custom dye calibration as normal, using 20 µL.

What dyes can I use on the QuantStudio 12K Flex Real-Time PCR System?

The QuantStudio 12K Flex Real-Time PCR System features a 6-color filter set that supports all Life Technologies dyes. Custom dyes that excite between 455 and 672 nm and read between 505 and 723 nm can also be used, although you will have to calibrate the system first for any new dye.

What are the specifications for the QuantStudio 12K Flex Real-Time PCR System?

You can find information on the QuantStudio 12K Flex instrument specifications here:
http://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/real-time-pcr-instruments.html

Can I reuse my calibration plates for the QuantStudio 12K Flex Real-Time PCR System?

All the calibration plates can be stored and reused three times for up to 6 months after you first open them, so make sure to return them to their original packaging and return them to -20 degrees C storage until the next use. (Note: Due to the small volumes, calibration array cards cannot be reused. Please discard them after use.) If needed, you can make your own background plate using deionized water. Please follow the directions in the QuantStudio 12K Flex Real-Time PCR System Maintenance and Administration Guide (Appendix C) for more details at http://www.thermofisher.com/content/dam/LifeTech/migration/files/pcr/pdfs.par.4982.file.dat/4470689b.pdf.

What are the uniformity and normalization calibrations for in the QuantStudio 12K Flex Real-Time PCR System?

The uniformity calibration generates data that allows the software to compensate for the physical effects of the QuantStudio 12K Flex Real-Time PCR System filters using the ROI plate. The normalization calibration generates factors that the software uses when comparing data from multiple QuantStudio instruments within a study. There are 2 normalization plates provided in the calibration kit, one containing FAM/ROX dye and the other with VIC/ROX dye.

When do I need to calibrate the QuantStudio 12K Flex Real-Time PCR System?

The maintenance schedule can be found in the QuantStudio 12K Flex Maintenance and Administration Guide (Chapter 2). In short, the background calibration should be performed monthly, while all other calibrations should be performed annually.

How do I import sample names into the QuantStudio 12K Flex system?

The QuantStudio 12K Flex software can import sample files to populate sample information into an open experiment. There are many ways to import sample names into the software. You can import samples from a text file, a plate setup file, or a sample definition file. See p.206 of the Maintenance and Administartion User Guide, or search the software menu for "import samples'' for more directions. Please visit the QuantStudio 12K Flex Support Center (https://www.thermofisher.com/us/en/home/technical-resources/technical-reference-library/real-time-digital-PCR-instruments-support-center/quantstudio-12k-flex-real-time-pcr-system-support.html) to find the Maintenance Guide.

What are the electrical requirements of the QuantStudio 12K Flex system?

Please see p.22 of the QuantStudio Maintenance and Administration Guide for electrical requirements. Please visit the QuantStudio 12K Flex Support Center (https://www.thermofisher.com/us/en/home/technical-resources/technical-reference-library/real-time-digital-PCR-instruments-support-center/quantstudio-12k-flex-real-time-pcr-system-support.html) to find the Maintenance Guide.

Can I incorporate the QuantStudio 12K Flex system into a LIMS system?

The QuantStudio 12K Flex software supports several import file formats that can be used to automate experiment creation and assay/sample data import. The files can be used with the command-line application or the QuantStudio 12K Flex software application programming interface (API) to integrate the QuantStudio 12K Flex system into a laboratory information management system (LIMS). Search for Cat. No. 4470689 on the website to find the Maintenance Guide for more details.

How often should I calibrate the QuantStudio 12K Flex system?

The recommended maintenance schedule can be found on p.82 of the Maintenance and Administration User Guide. Search for Cat. No. 4470689 on the website to find the Maintenance Guide.

Does the QuantStudio 12K Flex software have a study option?

No. Relative quantitation data from the QuantStudio 12K Flex system is meant to be analyzed in ExpressionSuite software. ExpressionSuite software is a free analysis program available on the Technical Resources-->Software, Patches & Updates page.

Can I change the default block type for the QuantStudio 12K Flex system?

Yes. Go to Tools > Preferences > 'non-openarray' tab and change the dropdown for block type as needed.

Can I see the amplifcation plot on the instrument touchscreen?

Yes, on the QuantStudio 12K Flex system you can see the real time amplification plots on the touchscreen. On other systems, you will only be able to see the amplification plots in real-time on the connected computer.

What is the normalization calibration for in the ViiA 7 or QuantStudio 12K Flex systems?

The normalization calibration generates factors that the software uses when comparing data from multiple instruments.

What is the life span of the QuantStudio 12K Flex system light source?

The light source is a white LED. Depending on usage, it should last up to 5 years.

Can I use caps on the 96-well plates on the QuantStudio 12K Flex system?

Yes. We recommend that you use Optical Flat caps, such as Cat. No. N8010535. Use of domed caps may damage the heated cover.

Is there a way to get the instrument serial number from the QuantStudio 12K Flex system software?

Yes, from the instrument touchscreen, go to Settings > About the instrument. If you are using the instrument software (when connected) go to Instrument > Instrument Console > Instrument Properties menu.

Can I import a standard curve for my real-time data?

It is possible to import a standard curve if you have either the ViiA 7 or QuantStudio 12K Flex systems. In the ViiA 7 or QuantStudio instrument software, open the data file and go to 'Analysis Settings'. Click on the 'Standard Curve Settings' tab. You will see an option to 'Use a standard curve imported from another experiment'. Choose this, then click on "Import". Browse to the proper data file and click "Select".

On the QuantStudio 12K Flex system, do I have to change the heated cover when changing the block?

Yes. You will need to match all the accessories (heated cover and plate adapter) to the block you have installed. For the QuantStudio 12K Flex system, the 96-Standard and 96-Fast blocks use the same heated cover. The 384-well and TaqMan Array Card block also use the same heated cover. The OpenArray block uses it own separate heated cover.

What real-time instruments can be run independently from a computer?

The StepOne, StepOnePlus, ViiA 7, and QuantStudio 12K Flex systems can all be run directly from the instrument touchscreen and don't need a separate computer to operate them.

What is the difference between continuous melt curves and step and hold melt curves on the Applied Biosystems Real-Time PCR Instruments?

Continuous melt curve: increases the temperature by the ramp increment (degrees C/sec) so that the temperature will change at a constant rate set in the protocol.
Step and Hold melt curve: increases the temperature by the ramp increment (degrees C) and then holds at that temperature for the time specified by the user.

The amount of data collected will depend on the filter(s) selected. The more filters you use, the longer the acquisition will take and the fewer data points will be collected.

I recently switched the block in my real-time PCR instrument. Now it shows the status as 'not calibrated' for all the calibration plates, even though my calibrations are current. What is causing this issue?

The issue might be due to the block not being properly seated in the instrument. Please follow the steps below:

  1. Remove the block from the instrument.
  2. Reinsert the block into the instrument.


If the problem persists after reseating the block, please contact our Technical Support team for further assistance.

Can I import a standard curve for my real-time PCR data into the Design & Analysis Software (v2.x)?

It is possible to import a standard curve into the Design & Analysis Software (v2.x) if you are analyzing results from one of the following Applied Biosystems real-time PCR instruments:
• 7900HT Fast Real-Time PCR System
• 7500/7500 Fast Real-Time PCR System
• StepOne/StepOne Plus Real-Time PCR System
• ViiA 7 Real Time PCR System
• QuantStudio 1 Real-Time PCR System
• QuantStudio 3 & 5 Real-Time PCR System
• QuantStudio 6 & 7 Flex Real-Time PCR System
• QuantStudio 6 & 7 Pro Real-Time PCR Systems
• QuantStudio 12K Flex Real-Time PCR system

Follow the instructions below to import a standard curve for analysis in the Design & Analysis Software (v2.x):
1. Open the data file in the Design & Analysis Software (v2.x).
2. Click on Actions.
3. Click on "Standard Curve Analysis Setting".
4. Select "External Standard Curves" and click on "Import".
5. Browse to the proper .csv file and click "Open".
6. Click "Apply".

Can I import a standard curve for my real-time PCR data into the QuantStudio 12K Flex Software?

If you are analyzing results from the QuantStudio 12K Flex Real-Time PCR System, it is possible to import a standard curve into the QuantStudio 12K Flex Software following the instructions below:
1. Open the data file in the QuantStudio 12K Flex Software.
2. Go to Analysis Settings.
3. Click on the "Standard Curve Settings" tab.
4. Select the option to "Use a standard curve imported from another experiment" and click on "Import".
5. Browse to and highlight the proper data file and click "Select".
6. Click Apply Analysis Settings.

What is the difference between a Fast 96-well and standard 96-well block?

There are two main differences between a Fast 96-well and standard 96-well block for the Applied Biosystems real-time PCR systems:
• Reaction well volume: The Fast 96-well blocks have 0.1 mL reaction wells and are compatible with 0.1 mL plates and tubes. The standard 96-well blocks have 0.2 mL reaction wells and are compatible with 0.2 mL plates and tubes.
• Ramp rate: The fast 96-well blocks have a higher maximum ramp rate than standard blocks for the same real-time PCR system. However, both the fast 96-well and standard 96-well blocks are capable of running standard and fast chemistry.

Note: For newer instruments (QuantStudio 3 and 5 Real-Time PCR Systems and later) "0.1 mL" and "0.2 mL" are being used in place of "fast" and "standard" as designations for the two 96-well block formats.

How do I calibrate the QuantStudio 12K Flex Real-Time PCR instrument with Cy5 dye?

If you are running software v1.5.1 and below on the QuantStudio 12K Flex Real-Time PCR instrument and using a 384-well plate, you may reach out to our Specialty Oligos team at Specialty_Oligos@thermofisher.com to request Cy5 dye calibration oligos. Each tube is provided at 800 µL, 0.2 µM. For a 384-well plate at 10 µL/well, you will need to order two tubes, or at 20 µL/well, you will need to order three tubes. Please provide your shipping address to Specialty Oligos and they will provide you with a formal quote.

If you are using a 96-well block, the Cy5 dye plate contained in either 7500 Real Time PCR Systems Spectral Calibration Kit II (Cat. No. 4351151) (Standard, 0.2 mL) or 7500 Fast Real-Time PCR Systems Spectral Calibration Kit II (Cat. No. 4362201 (Fast, 0.1 mL) can be used in place of the Cy5 dye calibration oligos.

On the QuantStudio 12K Flex instrument, what does the error "Calibration file already exists" mean?

The calibration files are stored on the QuantStudio 12K Flex instrument; therefore, when you need to perform a new calibration, you will have to delete the old calibration files. From the instrument console, go to the QuantStudio 12K Flex instrument touch screen and select 'Collect Results'. You should then see a list of files stored on the instrument. Manually select a calibration file such as "Fast-96-cal" and press the 'Delete' button to remove the old calibration file. Occasionally, users cannot delete the file off the instrument and may encounter this error: "Some experiments did not collect results therefore cannot be deleted." For these situations, launch the software, go to 'maintenance', manage files, and download the calibration file. The status of the experiment should be displayed as 'complete'. Next, delete the old calibration file and rerun the calibration.

What type of Real-Time PCR (Sequence Detection System) information is available from Applied Biosystems?

Tutorials demonstrating introductions to Real-Time PCR, guidelines for designing real-time assays using Primer Express software, and various instrument set-ups can be found under Life Technologies University on our website. User bulletins, manuals and Product inserts, protocols can be found as downloadable pdf files on our Technical Resources page.

What qPCR instruments are compatible with Thermo Fisher Cloud?

All our real-time PCR instruments are compatible with Thermo Fisher Cloud Apps except the 7500 Fast instrument where the eds files alone are compatible with Thermo Fisher Cloud Apps.

How can I run multiple batches of pre-reads and post-reads in the Applied Biosystems QuantStudio 12K Flex Real-Time PCR instrument?

To perform pre-read and post-read, choose Get Plate IDs and input the setup file either with Genotyping.edt or spf for your runs.

- Check the box for “Include Pre-read” and uncheck “Include Amplification”.
- Edit the file location and the file name and start the run.
- After your pre-read run has ended, you can either keep the run console on to continue with the post-read, or you can close it and perform other pre-read runs.

If you closed the run console and wish to perform the post-read, put the OpenArray in the same position as it was when you did the pre-read and choose Get Plate IDs again. Select the eds file that you had performed pre-read as the setup file and start the run. Note: it is important here that you select the .eds file, NOT the .spf or .edt file.
If you have placed the array at the wrong position, the software will prompt you to select the correct position.

How can I get additional throughput for the Applied Biosystems QuantStudio 12K Flex OpenArray System?

Additional Applied Biosystems OpenArray plates can be run off-line using a flat-block thermal cycler. The two thermal cyclers we recommend are the Applied Biosystems GeneAmp PCR System 9700 and the Applied Biosystems ProFlex PCR System.

How do I get my log files from the Applied Biosystems QuantStudio 12K Flex Real-Time PCR instrument?

To get log files from the QuantStudio 12K Flex Real-Time PCR instrument,

1. Insert a USB cord into the instrument.
2. Follow the instructions in the “Tools” menu on the touch-screen.
3. Download all 5 log files at once to the USB.

To get log files from the QuantStudio 12K Flex Real-Time PCR software on the client computer,

1. Navigate to this path: C:\Applied Biosystems\AppData\QuantStudio 12K Flex Software\UserData\data\log
2. Zip the entire contents of this directory.
3. Navigate to this path: C:\Applied Biosystems\AppData\QuantStudio 12K Flex Software\MetaData\logs
4. Zip the entire contents of this directory.

I would like to calibrate my Applied Biosystems QuantStudio 12K Flex Real-Time PCR instrument with a custom dye, which buffer should I use?

Please ask your dye manufacturer for recommendations. If there is no specific recommendation available, try TE Buffer (pH 8.0) to start.