missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ QuantiGene™ Sample Processing Kit, blood samples
Description
This QuantiGene Sample Processing Kit for blood samples contains reagents and instructions for the preparation of cell lysates from whole blood samples for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA or DNA targets.
- Easy to follow instructions
- Preserves the integrity of the RNA/DNA without degradation
- Choose the method that matches your whole blood or dried blood spot sample
What you get
The kit contains our proprietary lysis mixture, proteinase K, and detailed instructions for preparing whole blood from heparin, citrate, or EDTA tubes, PAXgene blood RNA tubes, Tempus™ blood RNA tubes, or from dried blood spots prepared from standard anticoagulant venous blood or finger sticks.
Specifications
Specifications
| Content And Storage | • 50 mL Lysis Mixture, store at 15°C to 30°C • 3.25 mL Proteinase K (50 μg/μL), store at -20°C |
| Format | Kit |
| Sample Type | Blood |
| Product Line | QuantiGene |
| Product Type | Sample Processing Kit |
| Quantity | 10 Plates |
| Shipping Condition | Blue Ice |