missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ QuantiGene™ Plex Human JAK-STAT Signaling Pathway Panel, 23-plex

The QuantiGene Plex Human JAK-STAT Signaling Pathway Panel enables measurement of expression of relevant human genes associated with the JAK-STAT pathway through the analysis of 23 RNA targets and housekeeping genes using Luminex xMAP technology in combination with branched DNA signal amplification.

Supplier:  Invitrogen™ QGP323JAKSTAT

 View more versions of this product

Catalog No. 5000153561

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

The QuantiGene Plex Human JAK-STAT Signaling Pathway Panel, 23-plex, enables measurement of expression of relevant human genes associated with the JAK-STAT signaling pathway through the analysis of 23 RNA targets and housekeeping genes in a single well using Luminex xMAP technology in combination with branched DNA signal amplification. This panel contains key genes that are involved in signaling pathway dynamics that regulate immune response, cell growth, and apoptosis, helping enable insights in oncology, immunology, and inflammatory diseases research.

QuantiGene Plex panels are extensively tested for target combinability, interference, and cross-reactivity to provide the high levels of validation and precision.

This panel is used in combination with the QuantiGene Plex Assay Kit, which contains all buffers, plates, and detection reagents necessary to perform the assay in 96-well plates.

Features of QuantiGene Plex panels include:

  • True multiplexing—measure multiple genes of interest and housekeeping genes in the same well with no cross-reactivity
  • Accommodation of difficult sample types—works with degraded and cross-linked RNA in FFPE tissues and blood
  • Standardized platform—uses 96-well format compatible with Luminex 200, FLEXMAP 3D, and INTELLIFLEX systems (384-well format available on demand)
  • Simple workflow—employs an ELISA-like workflow with direct hybridization of transcripts to beads and transcript labeling (branched DNA signal amplification and PE readout)
  • Customize with validated targetsif alternative targets are needed in this panel, select from our large inventory of over 22,000 genes to create custom research panels
  • Rapid turnaround of this standard panel for fast delivery

Targets: STAT1, STAT3, STAT5A, STAT5B, STAT6, JAK1, JAK2, JAK3, TYK2, SRC, FYN, PIAS1, PIAS3, SOCS1, SOCS3, PTPN1, PTPN2, PTPN6, PTPN11, GAPDH, HPRT1, PPIB, GUSB

Panel and signal amplification technology

QuantiGene Plex assays are hybridization-based assays for the direct quantitation of RNA directly from cell lysates, tissue homogenates, or purified RNA, and they enable the measurement of multiple targets in the same well via multiplexing. QuantiGene Plex panels contain two components: a target-specific probe set and magnetic capture beads. The target-specific probes are used to capture the RNA of interest, and branched DNA technology then amplifies the fluorescent signals, which are read with a Luminex xMAP instrument. The intensity of the fluorescent signal is directly proportional to the amount of target mRNA present in the sample. This allows for (relative) quantification of gene expression and, with this specific panel, enables human immune profiling. Detailed and step-by-step data analysis is facilitated by the QuantiGene Plex data analysis app.

Applications

QuantiGene Plex assay research applications include compound screening of drug candidates, biomarker verification, siRNA knockdown efficiency determination, prospective and/or retrospective analysis of clinical samples, microarray validation, predictive toxicology research, and translocation/fusion gene detection. In general, any multiplex gene expression analysis can be done with QuantiGene Plex assays, independent of species or sample type.

Specifications

Chemical Identifiers

96-well
Luminex Instrument
QuantiGene™ Plex
Wet Ice
Fluorescence
Human
QuantiGene Gene Expression Assay

Target-specific probe set and magnetic capture beads (premixed and ready to use).

Store probe set and capture beads at 4°C.

Do not freeze the capture beads, as they can be damaged if frozen.

Multiplex Panel
Purified RNA, cell/blood/FFPE lysates, tissue homogenates
Cell Signaling

Specifications

96-well
Luminex Instrument
QuantiGene™ Plex
10 Plates
Purified RNA, cell/blood/FFPE lysates, tissue homogenates
Cell Signaling
QuantiGene Gene Expression Assay

Target-specific probe set and magnetic capture beads (premixed and ready to use).

Store probe set and capture beads at 4°C.

Do not freeze the capture beads, as they can be damaged if frozen.

Multiplex Panel
Wet Ice
Fluorescence
Human
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.