missing translation for 'onlineSavingsMsg'
Learn More

Promega QuantiFluor™ ssDNA System

Catalog No. PRE3190
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRE3190 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRE3190 Supplier Promega Supplier No. E3190
Only null left

Includes

  • QuantiFluor ssDNA Dye, 200X
  • ssDNA Standard
  • 20X TE Buffer, pH 7.5

The QuantiFluor ssDNA System contains a fluorescent dye that enables sensitive quantitation of small amounts of single-stranded (ssDNA) in solution.

  • Sensitive Quantitation of Single-Stranded DNA
  • Requires less DNA than spectrophotometry

Specifications

Content And Storage -30°C to -10°C
Quantity 1 mL