Learn More
Promega QuantiFluor™ ONE dsDNA System
The QuantiFluor(R) ONE dsDNA System enables sensitive quantitation of small amounts of double-stranded DNA (dsDNA). The system contains a fluorescent dsDNA-binding dye (504nm[Ex]/531nm[Em]) developed for use in an add-and-read format for dye and standard.
$169.97 - $540.21
Specifications
| Content And Storage | Varies Among Components |
|---|
Includes: QuantiFluor ONE dsDNA Dye
QuantiFluor ONE Lambda DNA
1X TE Buffer, pH 7.5
Description
- Fluorescent Quantitation of dsDNA with Add-and-Read Convenience
- Specific quantitation of dsDNA
- Increased sensitivity over absorbance at 260nm for low-concentration samples
- Simple to use and automation-friendly
Specifications
| Varies Among Components |