Learn More
Promega QuantiFluor™ dsDNA Sample Kit DFS Item
The QuantiFluor dsDNA System contains a fluorescent DNA-binding dye that enables sensitive quantitation of small amounts of double-stranded DNA (dsDNA) in solution. The dye shows minimal binding to single stranded DNA (ssDNA) and RNA.
Supplier:  Promega E2671
Catalog No. PRE2671
Only null left
Includes:
  • QuantiFluor dsDNA Dye, 200X
  • Lambda DNA Standard
  • 20X TE Buffer, pH 7.5
Description
Description
  • Sensitive Quantitation of Double-Stranded DNA in Solution
  • Specific quantitation of dsDNA
Specifications
Specifications
-30°C to +10°C
1/Ea.
Product Suggestions
Product Suggestions