missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Quant-iT™ PicoGreen™ dsDNA Assay Kits and dsDNA Reagents
Description
Achieve precise and accurate dsDNA concentration measurements over a broad dynamic range with the Quant-iT Picogreen dsDNA Assay kits and dsDNA reagents. Picogreen assays are compatible with most microplate readers and fluorometers.
Rapidly detect as little as 0.25 pg/uL of dsDNA, even in the presence of ssDNA, RNA, and free nucleotides, using the PicoGreen dsDNA quantitation assay and reagent. The assay is linear over three orders of magnitude and has little sequence dependence, allowing you to accurately measure DNA from many sources, including genomic DNA, viral DNA, miniprep DNA, or PCR amplification products.
PicoGreen dsDNA quantitation reagents are
• Significantly more sensitive than UV absorbance readings, saving precious sample amounts
• Specific for dsDNA in the presence of RNA
• Easy to use—just add the dye working solution to the sample, incubate five minutes, and read
• Suitable for use in 96- and 384-well plate formats
• Compatible with most fluorescence-based microplate readers and fluorometers
• Picogreen aproximate fluorescence absorption/emission maxima is 502/523 nm, bound to dsDNA
Applications
The PicoGreen dsDNA quantitation reagent and kits are ideal for PCR-based assays, microarray samples, DNA damage assays, enzyme activity assays, genomic DNA quantitation, measuring dsDNA in complex mixtures, and viral DNA quantitation.
PicoGreen dsDNA quantitation reagents are
• Significantly more sensitive than UV absorbance readings, saving precious sample amounts
• Specific for dsDNA in the presence of RNA
• Easy to use—just add the dye working solution to the sample, incubate five minutes, and read
• Suitable for use in 96- and 384-well plate formats
• Compatible with most fluorescence-based microplate readers and fluorometers
• Picogreen aproximate fluorescence absorption/emission maxima is 502/523 nm, bound to dsDNA
Applications
The PicoGreen dsDNA quantitation reagent and kits are ideal for PCR-based assays, microarray samples, DNA damage assays, enzyme activity assays, genomic DNA quantitation, measuring dsDNA in complex mixtures, and viral DNA quantitation.
Specifications
Specifications
| Detection Method | Fluorescence |
| Excitation/Emission | 500/525 |
| For Use With (Equipment) | Microplate Reader |
| No. of Reactions | 200 Assays (2 mL assay volume) |
| Packaging Type | 1 x 1 mL Bottle |
| Quantitation Range | 50 pg to 2 μg |
| Product Line | PICOGREEN, Quant-iT |
| Product Type | dsDNA Assay Kit |
| Quantity | 1 mL kit |
| Shipping Condition | Room Temperature |