missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Quant-iT™ dsDNA Assay Kits, high sensitivity (HS) and broad range (BR)
GREENER_CHOICE

For highly selective quantitation of double-stranded DNA over RNA, the Quant-iT dsDNA Assay Kits, high sensitivity and broad range, produce fluorescence signals that are linear in the ranges of 0.2-100 ng and 2-1000 ng of DNA, respectively.

$960.00 - $972.00

Specifications

Detection Method Fluorescence
For Use With (Equipment) Microplate Reader
No. of Reactions 1000 Assays (200 μL assay volume)
Product Line Quant-iT
Quantity 1 kit
View More Specs

Products 2
Catalog Number Mfr. No. Assay Quantitation Range Price Quantity & Availability  
Catalog Number Mfr. No. Assay Quantitation Range Price Quantity & Availability  
Q33120
GREENER_CHOICE
Encompass
View Documents Promotion Details
Invitrogen™
Q33120
dsDNA Quantitation, HS 0.2 to 100 ng
Each for $972.00
Only null left
 
Q33130
Encompass
View Documents Promotion Details
Invitrogen™
Q33130
dsDNA Quantitation, BR 4 to 1000 ng
Each for $960.00
Only null left
 
Description

Description

Perform dsDNA quantification easily and quickly with Quant-iT dsDNA assay kits. Both the Quant-iT High-Sensitivity dsDNA Assay Kit and the Quant-iT Broad-Range dsDNA Assay Kit provide concentrated assay reagent, dilution buffer, and pre-diluted DNA standards. These DNA assay kits are highly selective for double-stranded DNA over RNA, and, in the ranges of 0.2–100 ng DNA (for the HS dsDNA Assay Kit) and 4–1000 ng DNA (for the BR dsDNA Assay Kit), the fluorescence signal is linear.

The Quant-iT dsDNA High-Sensitivity Assay Kit and the Quant-iT dsDNA Broad-Range Assay Kit make DNA quantification easy and accurate. The kits provide concentrated assay reagent, dilution buffer, and prediluted DNA standards. Simply dilute the reagent 1:200, load 200 μL into the wells of a microplate, add 1–20 μL sample, mix, then read the fluorescence.

The assays are highly selective for double-stranded DNA over RNA, and, in the range of 0.2–100 ng for the HS assay or 4–1000 ng for the BR assay, the fluorescence signal is linear with DNA. The assays are performed at room temperature, and the signal is stable for three hours. Common contaminants such as salts, solvents, detergents, and protein are well tolerated in these assays.

Specifications

Specifications

Fluorescence
1000 Assays (200 μL assay volume)
1 kit
Microplate Reader
Quant-iT
Room Temperature
Videos
SDS
Documents

Documents