missing translation for 'onlineSavingsMsg'
Learn More

Quality Control Standards 23 For ICP, SPEX CertiPrep™

Catalog No. QC23
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QC23 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. QC23 Supplier SPEX CertiPrep Group LLC Supplier No. QC23

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ag1000μg/mL
Al1000μg/mL
B1000μg/mL
Ba1000μg/mL
Bi1000μg/mL
Ca1000μg/mL
Cd1000μg/mL
Co1000μg/mL
Cr1000μg/mL
Cu1000μg/mL
Fe1000μg/mL
Ga1000μg/mL
In1000μg/mL
K1000μg/mL
Li1000μg/mL
Mg1000μg/mL
Mn1000μg/mL
Na1000μg/mL
Ni1000μg/mL
Pb1000μg/mL
Sr1000μg/mL
Tl1000μg/mL
Zn1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Quality Control Standard 23
Name Note Standard 23
Packaging Triple-leached LDPE bottle
CAS 7697-37-2, 74-97-5, 7439-96-5, 7439-92-1, 7440-50-8
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 10.0, 90.0
Analyte or Component Names Ag,Al,B
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Element Ag, Al, B
Solvent or Matrix 10% HNO3
Quantity 125 mL
Grade Assurance
Show More Show Less

Product intended for laboratory use only