Learn More
MilliporeSigma™ Qualification Workbook
Provides instructions on performing system qual and completing qual forms
Supplier:  MilliporeSigma™ VLXSX005Y
 View more versions of this product
Catalog No. VLXSX005Y
Only null left
Explore available promotions
Includes:
IQ and OQ protocols, recommended PQ protocols and annual system maintenance procedures
Description
Description
  • For use with Milli-RO, Milli-RX, Milli-Q, RiOs, SDS, Elix, Direct-Q systems
Specifications
Specifications
Elix 5 System
Qualification Workbook
Provides instructions on performing system qual and completing qual forms
IQ and OQ protocols, recommended PQ protocols and annual system maintenance procedures
Product Suggestions
Product Suggestions