Learn More
MilliporeSigma™ Qualification Workbook
Provides instructions on performing system qual and completing qual forms
$2,650.00
Specifications
| Product Type | Qualification Workbook |
|---|---|
| For Use With (Application) | Provides instructions on performing system qual and completing qual forms |
| Includes | IQ and OQ protocols, recommended PQ protocols and annual system maintenance procedures |
Description
- For use with Milli-RO, Milli-RX, Milli-Q, RiOs, SDS, Elix, Direct-Q systems
Specifications
| Qualification Workbook | |
| Provides instructions on performing system qual and completing qual forms | |
| IQ and OQ protocols, recommended PQ protocols and annual system maintenance procedures |